Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569980_m1
          See other PLA2G2C GE Assays ›
          Gene Symbol:
          Pla2g2c
          Gene Name:
          phospholipase A2, group IIC
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 157163646 - 157187654 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569980_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019202.2 NP_062075.1 3 - 4 457 84
          XM_006239142.2 XP_006239204.1 3 - 4 475 84
          XM_006239143.2 XP_006239205.1 2 - 3 236 84
          XM_008764212.1 XP_008762434.1 3 - 4 550 84
          XM_008764213.1 XP_008762435.1 3 - 4 328 84
          XM_008764214.1 XP_008762436.1 3 - 4 1999 84
          GenBank mRNA BC097325.1 - 3 - 4 457 7456563 84
          U07798.1 - 3 - 4 784 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001757.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g2c

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid metabolic process
          lipid catabolic process
          phospholipase A2 activity
          calcium ion binding
          calcium-dependent phospholipase A2 activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P048668 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P760354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87506 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P760354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P760354 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P760354 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048668 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P760354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19335 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873328 - -
          Affymetrix Rat Gene 1.0 ST Array 10873329 - -
          Affymetrix Rat Gene 1.0 ST Array 10873332 - -
          Affymetrix Rat Gene 1.0 ST Array 10873330 - -
          Affymetrix Rat Gene 1.0 ST Array 10873334 - -
          Affymetrix Rat Gene 1.0 ST Array 10873335 - -
          Affymetrix Rat Gene 1.0 ST Array 10873331 - -
          Affymetrix Rat Gene 1.0 ST Array 10873333 - -
          Affymetrix Rat Gene 1.1 ST Array 10873327 - -
          Rat Genome 230 2.0 Array 1368594_PM_at - -
          Rat Genome 230 2.0 Array 1368594_at - -
          Affymetrix HT Rat Focus Array 1368594_PM_at - -
          Affymetrix HT Rat Focus Array 1368594_at - -
          Affymetrix Rat Clariom S Assay PSR0500013059.rn.1 TC0500001757.rn.1 RTA TC
          Rat Expression Set 230 1368594_PM_at - -
          Rat Expression Set 230 1368594_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873327 - -
          Rat Genome U34 Set U07798UTR#1_at - -
          Rat Genome U34 Set U07798cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21678825 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360652 - -
          RatRef-12 Expression BeadChip ILMN_49009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline