Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570056_m1
          See other RASGRP1 GE Assays ›
          Gene Symbol:
          Rasgrp1
          Gene Name:
          RAS guanyl releasing protein 1 (calcium and DAG-regulated)
          Gene Aliases:
          Rasgrp
          Chromosome Location:
          Chr.3: 108984029 - 109044420 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasgrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019211.2 NP_062084.2 9 - 10 1386 103
          GenBank mRNA AF060819.1 - 9 - 10 1248 103
          AF081196.1 - 9 - 10 1386 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003941.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasgrp1

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          inflammatory response to antigenic stimulus
          Ras protein signal transduction
          regulation of phosphatidylinositol 3-kinase signaling
          cell differentiation
          secretory granule localization
          mast cell degranulation
          vesicle transport along microtubule
          calcium ion binding
          Rap guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30512 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11980 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11980 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35062 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4276 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848404 - -
          Affymetrix Rat Gene 1.0 ST Array 10848408 - -
          Affymetrix Rat Gene 1.0 ST Array 10848406 - -
          Affymetrix Rat Gene 1.0 ST Array 10848399 - -
          Affymetrix Rat Gene 1.0 ST Array 10848400 - -
          Affymetrix Rat Gene 1.0 ST Array 10848397 - -
          Affymetrix Rat Gene 1.0 ST Array 10848413 - -
          Affymetrix Rat Gene 1.0 ST Array 10848410 - -
          Affymetrix Rat Gene 1.0 ST Array 10848403 - -
          Affymetrix Rat Gene 1.0 ST Array 10848412 - -
          Affymetrix Rat Gene 1.0 ST Array 10848405 - -
          Affymetrix Rat Gene 1.0 ST Array 10848402 - -
          Affymetrix Rat Gene 1.0 ST Array 10848407 - -
          Affymetrix Rat Gene 1.0 ST Array 10848411 - -
          Affymetrix Rat Gene 1.0 ST Array 10848394 - -
          Affymetrix Rat Gene 1.0 ST Array 10848401 - -
          Affymetrix Rat Gene 1.0 ST Array 10848398 - -
          Affymetrix Rat Gene 1.0 ST Array 10848409 - -
          Affymetrix Rat Gene 1.1 ST Array 10848393 - -
          Rat Genome 230 2.0 Array 1369130_PM_at - -
          Rat Genome 230 2.0 Array 1369129_at - -
          Rat Genome 230 2.0 Array 1369129_PM_at - -
          Rat Genome 230 2.0 Array 1369130_at - -
          Affymetrix HT Rat Focus Array 1369130_at - -
          Affymetrix HT Rat Focus Array 1369129_PM_at - -
          Affymetrix HT Rat Focus Array 1369129_at - -
          Affymetrix HT Rat Focus Array 1369130_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300029844.rn.1 TC0300003941.rn.1 RTA TC
          Rat Expression Set 230 1369130_PM_at - -
          Rat Expression Set 230 1369130_at - -
          Rat Expression Set 230 1369129_at - -
          Rat Expression Set 230 1369129_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848393 - -
          Rat Genome U34 Set AF060819_s_at - -
          Rat Genome U34 Set AF081196_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20845594 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357641 - -
          RatRef-12 Expression BeadChip ILMN_53715 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline