Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570376_m1
          See other PCSK7 GE Assays ›
          Gene Symbol:
          Pcsk7
          Gene Name:
          proprotein convertase subtilisin/kexin type 7
          Gene Aliases:
          PC7
          Chromosome Location:
          Chr.8: 50200069 - 50222655 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570376_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcsk7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019246.1 NP_062119.1 9 - 10 1399 73
          GenBank mRNA U36580.1 - 9 - 10 1399 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000607.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcsk7

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing
          serine-type endopeptidase activity
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P461201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 750 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461201 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P461201 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461201 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700372036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3863 - -
          Agilent Rat cDNA Microarray (G4105A) 700372036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909713 - -
          Affymetrix Rat Gene 1.0 ST Array 10909719 - -
          Affymetrix Rat Gene 1.0 ST Array 10909709 - -
          Affymetrix Rat Gene 1.0 ST Array 10909706 - -
          Affymetrix Rat Gene 1.0 ST Array 10909708 - -
          Affymetrix Rat Gene 1.0 ST Array 10909705 - -
          Affymetrix Rat Gene 1.0 ST Array 10909714 - -
          Affymetrix Rat Gene 1.0 ST Array 10909711 - -
          Affymetrix Rat Gene 1.0 ST Array 10909715 - -
          Affymetrix Rat Gene 1.0 ST Array 10909712 - -
          Affymetrix Rat Gene 1.0 ST Array 10909718 - -
          Affymetrix Rat Gene 1.0 ST Array 10909717 - -
          Affymetrix Rat Gene 1.0 ST Array 10909707 - -
          Affymetrix Rat Gene 1.0 ST Array 10909720 - -
          Affymetrix Rat Gene 1.0 ST Array 10909710 - -
          Affymetrix Rat Gene 1.0 ST Array 10909721 - -
          Affymetrix Rat Gene 1.0 ST Array 10909716 - -
          Affymetrix Rat Gene 1.1 ST Array 10909704 - -
          Rat Genome 230 2.0 Array 1367886_PM_at - -
          Rat Genome 230 2.0 Array 1367886_at - -
          Affymetrix HT Rat Focus Array 1367886_at - -
          Affymetrix HT Rat Focus Array 1367886_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003953.rn.1 TC0800000607.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003954.rn.1 TC0800000607.rn.1 RTA TC
          Rat Expression Set 230 1367886_at - -
          Rat Expression Set 230 1367886_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909704 - -
          Rat Genome U34 Set U36580_at - -
          Rat Genome U34 Set rc_AI073232_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22293413 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371061 - -
          RatRef-12 Expression BeadChip ILMN_48909 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline