Search
Search
| Gene Symbol: Srsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195505.1 | NP_001182434.1 | 393 | 94 | ||
| NM_001195506.1 | NP_001182435.1 | 264 | 94 | |||
| NM_019257.3 | NP_062130.2 | 268 | 94 | |||
| XM_008764878.1 | XP_008763100.1 | 2 - 3 | 444 | 94 | ||
| XM_008764879.1 | XP_008763101.1 | 2 - 3 | 315 | 94 | ||
| GenBank mRNA | AF020683.1 | - | 2 - 3 | 330 | 94 | |
| BC058479.1 | - | 262 | 6920786 | 94 | ||
| FM049719.1 | - | 2 - 3 | 356 | 94 | ||
| FM059586.1 | - | 2 - 3 | 242 | 94 | ||
| FM081441.1 | - | 2 - 3 | 268 | 94 | ||
| L13635.1 | - | 364 | 94 | |||
Set Membership: |
|