Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570487_m1
          See other SCN11A GE Assays ›
          Gene Symbol:
          Scn11a
          Gene Name:
          sodium channel, voltage-gated, type XI, alpha subunit
          Gene Aliases:
          NaN
          Chromosome Location:
          Chr.8: 128450793 - 128521129 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570487_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019265.2 NP_062138.1 15 - 16 2420 110
          XM_008766674.1 XP_008764896.1 15 - 16 2440 110
          GenBank mRNA AF059030.2 - 15 - 16 2420 110
          AJ237852.1 - 14 - 15 2369 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003552.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn11a

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          neuronal action potential
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          regulation of sensory perception of pain
          membrane depolarization during action potential
          voltage-gated sodium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102979 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002191 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002191 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25150 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20372 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920939 - -
          Affymetrix Rat Gene 1.0 ST Array 10920949 - -
          Affymetrix Rat Gene 1.0 ST Array 10920951 - -
          Affymetrix Rat Gene 1.0 ST Array 10920936 - -
          Affymetrix Rat Gene 1.0 ST Array 10920942 - -
          Affymetrix Rat Gene 1.0 ST Array 10920941 - -
          Affymetrix Rat Gene 1.0 ST Array 10920955 - -
          Affymetrix Rat Gene 1.0 ST Array 10920938 - -
          Affymetrix Rat Gene 1.0 ST Array 10920940 - -
          Affymetrix Rat Gene 1.0 ST Array 10920948 - -
          Affymetrix Rat Gene 1.0 ST Array 10920937 - -
          Affymetrix Rat Gene 1.0 ST Array 10920947 - -
          Affymetrix Rat Gene 1.0 ST Array 10920943 - -
          Affymetrix Rat Gene 1.0 ST Array 10920944 - -
          Affymetrix Rat Gene 1.0 ST Array 10920952 - -
          Affymetrix Rat Gene 1.0 ST Array 10920934 - -
          Affymetrix Rat Gene 1.0 ST Array 10920932 - -
          Affymetrix Rat Gene 1.0 ST Array 10920935 - -
          Affymetrix Rat Gene 1.0 ST Array 10920954 - -
          Affymetrix Rat Gene 1.0 ST Array 10920945 - -
          Affymetrix Rat Gene 1.0 ST Array 10920930 - -
          Affymetrix Rat Gene 1.0 ST Array 10920946 - -
          Affymetrix Rat Gene 1.0 ST Array 10920931 - -
          Affymetrix Rat Gene 1.0 ST Array 10920950 - -
          Affymetrix Rat Gene 1.0 ST Array 10920953 - -
          Affymetrix Rat Gene 1.0 ST Array 10920933 - -
          Affymetrix Rat Gene 1.1 ST Array 10920929 - -
          Rat Genome 230 2.0 Array 1368768_PM_at - -
          Rat Genome 230 2.0 Array 1368768_at - -
          Affymetrix HT Rat Focus Array 1368768_at - -
          Affymetrix HT Rat Focus Array 1368768_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800028955.rn.1 TC0800003552.rn.1 RTA TC
          Rat Expression Set 230 1368768_PM_at - -
          Rat Expression Set 230 1368768_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920929 - -
          Rat Genome U34 Set AF059030_at - -
          Rat Genome U34 Set AF059030_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21291489 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364700 - -
          RatRef-12 Expression BeadChip ILMN_65195 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline