Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570571_m1
          See other KCNMB1 GE Assays ›
          Gene Symbol:
          Kcnmb1
          Gene Name:
          potassium channel subfamily M regulatory beta subunit 1
          Gene Aliases:
          BKbeta, BKbeta1, k(VCA)beta-1, slo-beta, slo-beta-1
          Chromosome Location:
          Chr.10: 18910811 - 18920125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570571_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnmb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019273.1 NP_062146.1 287 92
          GenBank mRNA AF020712.1 - 287 92

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF020712.1, NM_019273.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnmb1

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of calcium ion
          chemical synaptic transmission
          response to calcium ion
          potassium ion transmembrane transport
          calcium-activated potassium channel activity
          potassium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76503 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045750 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045750 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045750 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480135 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278569 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702133464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2570 - -
          Agilent Rat cDNA Microarray (G4105A) 702133464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732722 - -
          Affymetrix Rat Gene 1.0 ST Array 10732717 - -
          Affymetrix Rat Gene 1.0 ST Array 10732719 - -
          Affymetrix Rat Gene 1.0 ST Array 10732720 - -
          Affymetrix Rat Gene 1.0 ST Array 10732721 - -
          Affymetrix Rat Gene 1.0 ST Array 10732724 - -
          Affymetrix Rat Gene 1.0 ST Array 10732723 - -
          Affymetrix Rat Gene 1.0 ST Array 10732718 - -
          Affymetrix Rat Gene 1.1 ST Array 10732716 - -
          Rat Genome 230 2.0 Array 1369767_PM_a_at - -
          Rat Genome 230 2.0 Array 1369767_a_at - -
          Affymetrix HT Rat Focus Array 1369767_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369767_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000003215.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003206.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003216.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003212.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003210.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003214.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003207.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003211.rn.1 TC1000000337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003213.rn.1 TC1000000337.rn.1 RTA TC
          Rat Expression Set 230 1369767_a_at - -
          Rat Expression Set 230 1369767_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732716 - -
          Rat Genome U34 Set AF020712_at - -
          Rat Genome U34 Set AB010963_s_at - -
          Rat Genome U34 Set AF020712_at - -
          Rat Genome U34 Set AB010963_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22149628 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352186 - -
          RatRef-12 Expression BeadChip ILMN_68507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0