Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570576_m1
          See other COLQ GE Assays ›
          Gene Symbol:
          Colq
          Gene Name:
          collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 7626380 - 7681621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570576_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Colq
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019274.2 NP_062147.1 12 - 13 863 85
          GenBank mRNA AF007583.1 - 12 - 13 863 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002122.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Colq

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synaptic growth at neuromuscular junction
          neurotransmitter catabolic process
          skeletal muscle acetylcholine-gated channel clustering
          establishment of protein localization to membrane
          structural molecule activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142242 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142242 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142242 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P141051 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24830 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23313 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86019 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790246 - -
          Affymetrix Rat Gene 1.0 ST Array 10790245 - -
          Affymetrix Rat Gene 1.0 ST Array 10790251 - -
          Affymetrix Rat Gene 1.0 ST Array 10790250 - -
          Affymetrix Rat Gene 1.0 ST Array 10790244 - -
          Affymetrix Rat Gene 1.0 ST Array 10790247 - -
          Affymetrix Rat Gene 1.0 ST Array 10790254 - -
          Affymetrix Rat Gene 1.0 ST Array 10790242 - -
          Affymetrix Rat Gene 1.0 ST Array 10790253 - -
          Affymetrix Rat Gene 1.0 ST Array 10790248 - -
          Affymetrix Rat Gene 1.0 ST Array 10790255 - -
          Affymetrix Rat Gene 1.0 ST Array 10790240 - -
          Affymetrix Rat Gene 1.0 ST Array 10790239 - -
          Affymetrix Rat Gene 1.0 ST Array 10790252 - -
          Affymetrix Rat Gene 1.0 ST Array 10790243 - -
          Affymetrix Rat Gene 1.0 ST Array 10790256 - -
          Affymetrix Rat Gene 1.0 ST Array 10790249 - -
          Affymetrix Rat Gene 1.1 ST Array 10790238 - -
          Rat Genome 230 2.0 Array 1368475_at - -
          Rat Genome 230 2.0 Array 1368475_PM_at - -
          Affymetrix HT Rat Focus Array 1368475_PM_at - -
          Affymetrix HT Rat Focus Array 1368475_at - -
          Affymetrix Rat Clariom S Assay PSR1600016005.rn.1 TC1600002122.rn.1 RTA TC
          Rat Expression Set 230 1368475_PM_at - -
          Rat Expression Set 230 1368475_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790238 - -
          Rat Genome U34 Set AF007583_at - -
          Rat Genome U34 Set AF007583_at - -
          Rat Genome U34 Set AF007583_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616866 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353156 - -
          RatRef-12 Expression BeadChip ILMN_69132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline