Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570728_m1
          See other CDK1 GE Assays ›
          Gene Symbol:
          Cdk1
          Gene Name:
          cyclin-dependent kinase 1
          Gene Aliases:
          Cdc2, Cdc2a
          Chromosome Location:
          Chr.20: 20576343 - 20591510 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570728_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019296.1 NP_062169.1 7 - 8 891 76
          XM_006256353.2 XP_006256415.1 8 - 9 1018 76
          GenBank mRNA BC091549.1 - 8 - 9 976 7388690 76
          FQ227059.1 - 7 - 8 894 76
          X60767.1 - 7 - 8 891 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000542.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdk1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein complex assembly
          protein phosphorylation
          apoptotic process
          mitotic nuclear division
          mitotic G2 DNA damage checkpoint
          cell aging
          response to toxic substance
          response to organonitrogen compound
          positive regulation of gene expression
          response to organic cyclic compound
          response to amine
          response to activity
          histone phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          chromosome condensation
          epithelial cell differentiation
          organ regeneration
          positive regulation of protein import into nucleus, translocation
          protein localization to kinetochore
          response to drug
          response to hydrogen peroxide
          negative regulation of apoptotic process
          response to ethanol
          positive regulation of DNA replication
          positive regulation of mitotic cell cycle
          response to cadmium ion
          response to copper ion
          response to axon injury
          cell division
          ventricular cardiac muscle cell development
          positive regulation of cardiac muscle cell proliferation
          cellular response to hydrogen peroxide
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          cyclin binding
          Hsp70 protein binding
          histone kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P580844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P580844 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P580844 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P580844 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P580844 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700948589 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701313344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54637 - -
          Agilent Rat cDNA Microarray (G4105A) 701313344 - -
          Agilent Rat cDNA Microarray (G4105A) 700948589 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829764 - -
          Affymetrix Rat Gene 1.0 ST Array 10829765 - -
          Affymetrix Rat Gene 1.0 ST Array 10829763 - -
          Affymetrix Rat Gene 1.0 ST Array 10829762 - -
          Affymetrix Rat Gene 1.0 ST Array 10829770 - -
          Affymetrix Rat Gene 1.0 ST Array 10829766 - -
          Affymetrix Rat Gene 1.0 ST Array 10829769 - -
          Affymetrix Rat Gene 1.0 ST Array 10829768 - -
          Affymetrix Rat Gene 1.0 ST Array 10829767 - -
          Affymetrix Rat Gene 1.1 ST Array 10829761 - -
          Rat Genome 230 2.0 Array 1367776_at - -
          Rat Genome 230 2.0 Array 1367776_PM_at - -
          Affymetrix HT Rat Focus Array 1367776_at - -
          Affymetrix HT Rat Focus Array 1367776_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000005017.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005024.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005019.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005027.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005025.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005015.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005023.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005026.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005022.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005029.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005016.rn.1 TC2000000542.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005020.rn.1 TC2000000542.rn.1 RTA TC
          Rat Expression Set 230 1367776_at - -
          Rat Expression Set 230 1367776_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829761 - -
          Rat Genome U34 Set X60767mRNA_s_at - -
          Rat Genome U34 Set X60767mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21187195 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358490 - -
          RatRef-12 Expression BeadChip ILMN_58988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline