Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570936_m1
          See other NR4A2 GE Assays ›
          Gene Symbol:
          Nr4a2
          Gene Name:
          nuclear receptor subfamily 4, group A, member 2
          Gene Aliases:
          Nurr1
          Chromosome Location:
          Chr.3: 43110416 - 43119284 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570936_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr4a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019328.3 NP_062201.2 4 - 5 1267 104
          XM_006234201.2 XP_006234263.1 5 - 6 1485 104
          XM_008761861.1 XP_008760083.1 4 - 5 1401 104
          GenBank mRNA DQ092629.1 - 4 - 5 1198 104
          DQ092630.1 - 4 - 5 1034 104
          DQ092631.1 - 4 - 5 1212 104
          DQ092632.1 - 4 - 5 1202 104
          DQ092633.1 - 4 - 5 1034 104
          DQ092634.1 - 4 - 5 1034 104
          FQ222109.1 - 5 - 6 1507 104
          L08595.1 - 4 - 5 1264 104
          U01146.1 - 4 - 5 1198 104
          U72345.1 - 5 - 6 1491 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003140.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr4a2

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          response to amphetamine
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          nervous system development
          adult locomotory behavior
          post-embryonic development
          response to inorganic substance
          gene expression
          regulation of gene expression
          obsolete death
          response to insecticide
          central nervous system projection neuron axonogenesis
          central nervous system neuron differentiation
          habenula development
          neuron differentiation
          intracellular receptor signaling pathway
          cellular response to extracellular stimulus
          cellular response to oxidative stress
          regulation of dopamine metabolic process
          dopamine biosynthetic process
          dopamine metabolic process
          neuron maturation
          positive regulation of catalytic activity
          steroid hormone mediated signaling pathway
          negative regulation of neuron apoptotic process
          regulation of respiratory gaseous exchange
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          general adaptation syndrome
          dopaminergic neuron differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154952 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61554 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12023 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12023 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154952 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12023 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P522524 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16801 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845387 - -
          Affymetrix Rat Gene 1.0 ST Array 10845389 - -
          Affymetrix Rat Gene 1.0 ST Array 10845390 - -
          Affymetrix Rat Gene 1.0 ST Array 10845396 - -
          Affymetrix Rat Gene 1.0 ST Array 10845391 - -
          Affymetrix Rat Gene 1.0 ST Array 10845394 - -
          Affymetrix Rat Gene 1.0 ST Array 10845388 - -
          Affymetrix Rat Gene 1.0 ST Array 10845393 - -
          Affymetrix Rat Gene 1.0 ST Array 10845392 - -
          Affymetrix Rat Gene 1.0 ST Array 10845395 - -
          Affymetrix Rat Gene 1.0 ST Array 10845386 - -
          Affymetrix Rat Gene 1.0 ST Array 10845385 - -
          Affymetrix Rat Gene 1.1 ST Array 10845384 - -
          Rat Genome 230 2.0 Array 1369007_PM_at - -
          Rat Genome 230 2.0 Array 1387410_PM_at - -
          Rat Genome 230 2.0 Array 1369007_at - -
          Rat Genome 230 2.0 Array 1387410_at - -
          Affymetrix HT Rat Focus Array 1369007_PM_at - -
          Affymetrix HT Rat Focus Array 1387410_at - -
          Affymetrix HT Rat Focus Array 1369007_at - -
          Affymetrix HT Rat Focus Array 1387410_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300024288.rn.1 TC0300003140.rn.1 RTA TC
          Rat Expression Set 230 1387410_at - -
          Rat Expression Set 230 1369007_PM_at - -
          Rat Expression Set 230 1387410_PM_at - -
          Rat Expression Set 230 1369007_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845384 - -
          Rat Genome U34 Set L08595_at - -
          Rat Genome U34 Set U01146_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21630440 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355906 - -
          RatRef-12 Expression BeadChip ILMN_58852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline