Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570970_m1
          See other FURIN GE Assays ›
          Gene Symbol:
          Furin
          Gene Name:
          furin (paired basic amino acid cleaving enzyme)
          Gene Aliases:
          Pace, Pcsk3
          Chromosome Location:
          Chr.1: 142185092 - 142198166 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570970_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Furin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019331.1 NP_062204.1 8 - 9 1282 70
          XM_008759555.1 XP_008757777.1 8 - 9 1166 70
          XM_008759556.1 XP_008757778.1 8 - 9 1213 70
          GenBank mRNA X55660.1 - 8 - 9 1282 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005882.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Furin

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          signal peptide processing
          proteolysis
          aging
          cell proliferation
          regulation of signal transduction
          negative regulation of endopeptidase activity
          protein processing
          peptide hormone processing
          viral life cycle
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of low-density lipoprotein particle receptor catabolic process
          nerve growth factor production
          negative regulation of nerve growth factor production
          negative regulation of transforming growth factor beta1 production
          secretion by cell
          regulation of protein catabolic process
          peptide biosynthetic process
          regulation of low-density lipoprotein particle receptor biosynthetic process
          regulation of endopeptidase activity
          protease binding
          endopeptidase activity
          serine-type endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase activity
          peptide binding
          metal ion binding
          nerve growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P240953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160586 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160586 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P240953 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700482503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3518 - -
          Agilent Rat cDNA Microarray (G4105A) 700482503 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723121 - -
          Affymetrix Rat Gene 1.0 ST Array 10723127 - -
          Affymetrix Rat Gene 1.0 ST Array 10723116 - -
          Affymetrix Rat Gene 1.0 ST Array 10723129 - -
          Affymetrix Rat Gene 1.0 ST Array 10723120 - -
          Affymetrix Rat Gene 1.0 ST Array 10723124 - -
          Affymetrix Rat Gene 1.0 ST Array 10723126 - -
          Affymetrix Rat Gene 1.0 ST Array 10723117 - -
          Affymetrix Rat Gene 1.0 ST Array 10723118 - -
          Affymetrix Rat Gene 1.0 ST Array 10723115 - -
          Affymetrix Rat Gene 1.0 ST Array 10723130 - -
          Affymetrix Rat Gene 1.0 ST Array 10723122 - -
          Affymetrix Rat Gene 1.0 ST Array 10723119 - -
          Affymetrix Rat Gene 1.0 ST Array 10723128 - -
          Affymetrix Rat Gene 1.0 ST Array 10723125 - -
          Affymetrix Rat Gene 1.0 ST Array 10723123 - -
          Affymetrix Rat Gene 1.1 ST Array 10723114 - -
          Rat Genome 230 2.0 Array 1367778_at - -
          Rat Genome 230 2.0 Array 1367778_PM_at - -
          Affymetrix HT Rat Focus Array 1367778_at - -
          Affymetrix HT Rat Focus Array 1367778_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100046859.rn.1 TC0100005882.rn.1 RTA TC
          Rat Expression Set 230 1367778_PM_at - -
          Rat Expression Set 230 1367778_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723114 - -
          Rat Genome U34 Set X55660_g_at - -
          Rat Genome U34 Set X55660_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21550633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline