Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571015_m1
          See other EIF2AK2 GE Assays ›
          Gene Symbol:
          Eif2ak2
          Gene Name:
          eukaryotic translation initiation factor 2-alpha kinase 2
          Gene Aliases:
          Pkr, Prkr
          Chromosome Location:
          Chr.6: 1428845 - 1466193 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571015_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif2ak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019335.1 NP_062208.1 9 - 10 840 87
          XM_008764425.1 XP_008762647.1 8 - 9 1064 87
          XM_008764426.1 XP_008762648.1 9 - 10 906 87
          GenBank mRNA L29281.1 - 9 - 10 840 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001730.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif2ak2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          positive regulation of cytokine production
          transcription, DNA-templated
          translation
          protein phosphorylation
          response to mechanical stimulus
          response to virus
          response to toxic substance
          response to organic substance
          negative regulation of translation
          peptidyl-tyrosine phosphorylation
          endoplasmic reticulum unfolded protein response
          response to lipopolysaccharide
          positive regulation of chemokine production
          positive regulation of stress-activated MAPK cascade
          response to vitamin E
          negative regulation of osteoblast proliferation
          response to interferon-alpha
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          response to exogenous dsRNA
          negative regulation of viral genome replication
          innate immune response
          protein autophosphorylation
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          regulation of NLRP3 inflammasome complex assembly
          positive regulation of NIK/NF-kappaB signaling
          regulation of hematopoietic progenitor cell differentiation
          regulation of hematopoietic stem cell proliferation
          regulation of hematopoietic stem cell differentiation
          RNA binding
          double-stranded RNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P313825 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P467037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467037 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P313825 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P467037 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P313825 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P467037 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700812124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15253 - -
          Agilent Rat cDNA Microarray (G4105A) 700812124 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940690 - -
          Affymetrix Rat Gene 1.1 ST Array 10940690 - -
          Rat Genome 230 2.0 Array 1387242_PM_at - -
          Rat Genome 230 2.0 Array 1387242_at - -
          Affymetrix HT Rat Focus Array 1387242_at - -
          Affymetrix HT Rat Focus Array 1387242_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600012258.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012261.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012267.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012270.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012275.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012269.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012266.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012276.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012279.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012259.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012271.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012274.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012260.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012265.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012272.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012273.rn.1 TC0600001730.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012268.rn.1 TC0600001730.rn.1 RTA TC
          Rat Expression Set 230 1387242_at - -
          Rat Expression Set 230 1387242_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940690 - -
          Rat Genome U34 Set rc_AI013987_s_at - -
          Rat Genome U34 Set L29281_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21939859 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376267 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline