Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571027_m1
          See other RGS12 GE Assays ›
          Gene Symbol:
          Rgs12
          Gene Name:
          regulator of G-protein signaling 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 80975239 - 81069517 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571027_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019339.1 NP_062212.1 6 - 7 2524 93
          XM_006251351.2 XP_006251413.1 7 - 8 2656 93
          XM_006251352.2 XP_006251414.1 7 - 8 2660 93
          XM_006251353.2 XP_006251415.1 7 - 8 3180 93
          XM_008770350.1 XP_008768572.1 7 - 8 3193 93
          XM_008770351.1 XP_008768573.1 6 - 7 931 93
          GenBank mRNA AF035151.1 - 5 - 6 757 93
          FQ230909.1 - 5 - 6 661 93
          U92280.1 - 6 - 7 2524 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002081.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgs12

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular signal transduction
          termination of G-protein coupled receptor signaling pathway
          positive regulation of GTPase activity
          negative regulation of G-protein coupled receptor protein signaling pathway
          regulation of catalytic activity
          G-protein alpha-subunit binding
          receptor signaling protein activity
          GTPase activator activity
          protein binding
          GTPase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179621 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095900 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095900 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095900 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P532444 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702425431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22494 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2933 - -
          Agilent Rat cDNA Microarray (G4105A) 702425431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777561 - -
          Affymetrix Rat Gene 1.0 ST Array 10777566 - -
          Affymetrix Rat Gene 1.0 ST Array 10777559 - -
          Affymetrix Rat Gene 1.0 ST Array 10777554 - -
          Affymetrix Rat Gene 1.0 ST Array 10777560 - -
          Affymetrix Rat Gene 1.0 ST Array 10777555 - -
          Affymetrix Rat Gene 1.0 ST Array 10777563 - -
          Affymetrix Rat Gene 1.0 ST Array 10777553 - -
          Affymetrix Rat Gene 1.0 ST Array 10777557 - -
          Affymetrix Rat Gene 1.0 ST Array 10777565 - -
          Affymetrix Rat Gene 1.0 ST Array 10777550 - -
          Affymetrix Rat Gene 1.0 ST Array 10777564 - -
          Affymetrix Rat Gene 1.0 ST Array 10777549 - -
          Affymetrix Rat Gene 1.0 ST Array 10777556 - -
          Affymetrix Rat Gene 1.0 ST Array 10777552 - -
          Affymetrix Rat Gene 1.0 ST Array 10777551 - -
          Affymetrix Rat Gene 1.0 ST Array 10777562 - -
          Affymetrix Rat Gene 1.0 ST Array 10777558 - -
          Affymetrix Rat Gene 1.1 ST Array 10777547 - -
          Rat Genome 230 2.0 Array 1387066_a_at - -
          Rat Genome 230 2.0 Array 1387066_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387066_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387066_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400014815.rn.1 TC1400002081.rn.1 RTA TC
          Rat Expression Set 230 1387066_PM_a_at - -
          Rat Expression Set 230 1387066_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777547 - -
          Rat Genome U34 Set AF035151_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20774845 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358689 - -
          RatRef-12 Expression BeadChip ILMN_70072 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline