Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571208_g1
          See other ARC GE Assays ›
          Gene Symbol:
          Arc
          Gene Name:
          activity-regulated cytoskeleton-associated protein
          Gene Aliases:
          rg3.1
          Chromosome Location:
          Chr.7: 115907097 - 115911054 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571208_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019361.1 NP_062234.1 1 - 2 2008 119
          XM_008765590.1 XP_008763812.1 1 - 2 2519 119
          GenBank mRNA U19866.1 - 1 - 2 2008 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003333.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arc

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          cytoskeleton organization
          endoderm development
          learning
          anterior/posterior pattern specification
          cell migration
          regulation of cell morphogenesis
          regulation of neuronal synaptic plasticity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P118724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777168 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P118724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P118724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P118724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P118724 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702123535 - -
          Agilent Rat cDNA Microarray (G4105A) 702123535 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904514 - -
          Affymetrix Rat Gene 1.0 ST Array 10904513 - -
          Affymetrix Rat Gene 1.0 ST Array 10904512 - -
          Affymetrix Rat Gene 1.1 ST Array 10904511 - -
          Rat Genome 230 2.0 Array 1387068_at - -
          Rat Genome 230 2.0 Array 1387068_PM_at - -
          Affymetrix HT Rat Focus Array 1387068_at - -
          Affymetrix HT Rat Focus Array 1387068_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700025506.rn.1 TC0700003333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700025505.rn.1 TC0700003333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700025508.rn.1 TC0700003333.rn.1 RTA TC
          Rat Expression Set 230 1387068_PM_at - -
          Rat Expression Set 230 1387068_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904511 - -
          Rat Genome U34 Set U19866_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21357407 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039673 - -
          RatRef-12 Expression BeadChip ILMN_68689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline