Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571242_m1
          See other AOX1 GE Assays ›
          Gene Symbol:
          Aox1
          Gene Name:
          aldehyde oxidase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 64929679 - 65007872 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aox1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019363.3 NP_062236.2 26 - 27 3109 74
          GenBank mRNA AF110477.1 - 26 - 27 3126 74
          AF110478.1 - 26 - 27 3110 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000697.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aox1

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          drug metabolic process
          oxidation-reduction process
          aldehyde oxidase activity
          iron ion binding
          UDP-N-acetylmuramate dehydrogenase activity
          electron carrier activity
          molybdopterin cofactor binding
          flavin adenine dinucleotide binding
          NAD binding
          2 iron, 2 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091747 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091747 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1052607 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701645076 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701482962 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701619084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48219 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46557 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49896 - -
          Agilent Rat cDNA Microarray (G4105A) 701482962 - -
          Agilent Rat cDNA Microarray (G4105A) 701619084 - -
          Agilent Rat cDNA Microarray (G4105A) 701645076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923409 - -
          Affymetrix Rat Gene 1.0 ST Array 10923424 - -
          Affymetrix Rat Gene 1.0 ST Array 10923404 - -
          Affymetrix Rat Gene 1.0 ST Array 10923428 - -
          Affymetrix Rat Gene 1.0 ST Array 10923429 - -
          Affymetrix Rat Gene 1.0 ST Array 10923421 - -
          Affymetrix Rat Gene 1.0 ST Array 10923418 - -
          Affymetrix Rat Gene 1.0 ST Array 10923417 - -
          Affymetrix Rat Gene 1.0 ST Array 10923395 - -
          Affymetrix Rat Gene 1.0 ST Array 10923422 - -
          Affymetrix Rat Gene 1.0 ST Array 10923410 - -
          Affymetrix Rat Gene 1.0 ST Array 10923402 - -
          Affymetrix Rat Gene 1.0 ST Array 10923407 - -
          Affymetrix Rat Gene 1.0 ST Array 10923399 - -
          Affymetrix Rat Gene 1.0 ST Array 10923425 - -
          Affymetrix Rat Gene 1.0 ST Array 10923423 - -
          Affymetrix Rat Gene 1.0 ST Array 10923426 - -
          Affymetrix Rat Gene 1.0 ST Array 10923414 - -
          Affymetrix Rat Gene 1.0 ST Array 10923416 - -
          Affymetrix Rat Gene 1.0 ST Array 10923411 - -
          Affymetrix Rat Gene 1.0 ST Array 10923394 - -
          Affymetrix Rat Gene 1.0 ST Array 10923413 - -
          Affymetrix Rat Gene 1.0 ST Array 10923427 - -
          Affymetrix Rat Gene 1.0 ST Array 10923406 - -
          Affymetrix Rat Gene 1.0 ST Array 10923400 - -
          Affymetrix Rat Gene 1.0 ST Array 10923430 - -
          Affymetrix Rat Gene 1.0 ST Array 10923431 - -
          Affymetrix Rat Gene 1.0 ST Array 10923415 - -
          Affymetrix Rat Gene 1.0 ST Array 10923397 - -
          Affymetrix Rat Gene 1.0 ST Array 10923419 - -
          Affymetrix Rat Gene 1.0 ST Array 10923412 - -
          Affymetrix Rat Gene 1.0 ST Array 10923398 - -
          Affymetrix Rat Gene 1.0 ST Array 10923396 - -
          Affymetrix Rat Gene 1.0 ST Array 10923403 - -
          Affymetrix Rat Gene 1.0 ST Array 10923408 - -
          Affymetrix Rat Gene 1.0 ST Array 10923401 - -
          Affymetrix Rat Gene 1.0 ST Array 10923420 - -
          Affymetrix Rat Gene 1.1 ST Array 10923393 - -
          Rat Genome 230 2.0 Array 1387376_at - -
          Rat Genome 230 2.0 Array 1387376_PM_at - -
          Affymetrix HT Rat Focus Array 1387376_at - -
          Affymetrix HT Rat Focus Array 1387376_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900005484.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005491.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005506.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005508.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005510.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005533.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005492.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005504.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005507.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005525.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005528.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005494.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005502.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005518.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005529.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005534.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005485.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005486.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005488.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005497.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005519.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005512.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005532.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005489.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005522.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005524.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005527.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005530.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005523.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005536.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005487.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005490.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005493.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005495.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005505.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005516.rn.1 TC0900000697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005521.rn.1 TC0900000697.rn.1 RTA TC
          Rat Expression Set 230 1387376_PM_at - -
          Rat Expression Set 230 1387376_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923393 - -
          Rat Genome U34 Set rc_AA926200_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22308791 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370454 - -
          RatRef-12 Expression BeadChip ILMN_53249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline