Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571358_m1
          See other SEPT3 GE Assays ›
          Gene Symbol:
          Sept3
          Gene Name:
          septin 3
          Gene Aliases:
          Sep3
          Chromosome Location:
          Chr.7: 123506535 - 123526542 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019375.1 NP_062248.1 7 - 8 799 93
          GenBank mRNA AF111179.1 - 7 - 8 799 93
          AF111180.1 - 5 - 6 1206 93
          AF111181.1 - 7 - 8 798 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001525.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept3

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Process(es)

          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P645142 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16187 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P645142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P645142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P645142 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P645142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 898 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24418 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52489 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898120 - -
          Affymetrix Rat Gene 1.0 ST Array 10898122 - -
          Affymetrix Rat Gene 1.0 ST Array 10898126 - -
          Affymetrix Rat Gene 1.0 ST Array 10898123 - -
          Affymetrix Rat Gene 1.0 ST Array 10898127 - -
          Affymetrix Rat Gene 1.0 ST Array 10898125 - -
          Affymetrix Rat Gene 1.0 ST Array 10898129 - -
          Affymetrix Rat Gene 1.0 ST Array 10898124 - -
          Affymetrix Rat Gene 1.0 ST Array 10898128 - -
          Affymetrix Rat Gene 1.0 ST Array 10898131 - -
          Affymetrix Rat Gene 1.1 ST Array 10898119 - -
          Rat Genome 230 2.0 Array 1370116_PM_at - -
          Rat Genome 230 2.0 Array 1370116_at - -
          Affymetrix HT Rat Focus Array 1385489_PM_at - -
          Affymetrix HT Rat Focus Array 1370116_at - -
          Affymetrix HT Rat Focus Array 1370116_PM_at - -
          Affymetrix HT Rat Focus Array 1385489_at - -
          Affymetrix Rat Clariom S Assay PSR0700011685.rn.1 TC0700001525.rn.1 RTA TC
          Rat Expression Set 230 1370116_at - -
          Rat Expression Set 230 1370116_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898119 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21011049 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline