Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571431_m1
          See other GORASP1 GE Assays ›
          Gene Symbol:
          Gorasp1
          Gene Name:
          golgi reassembly stacking protein 1
          Gene Aliases:
          GRASP65
          Chromosome Location:
          Chr.8: 128610290 - 128622337 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571431_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gorasp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019385.2 NP_062258.2 3 - 4 370 80
          GenBank mRNA AF015264.2 - 3 - 4 347 80
          BC081809.1 - 3 - 4 364 80
          BC161850.1 - 3 - 4 370 8369057 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gorasp1

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi organization
          protein transport
          negative regulation of dendrite morphogenesis
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147973 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147973 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147973 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205913 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700369249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86275 - -
          Agilent Rat cDNA Microarray (G4105A) 700369249 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920962 - -
          Affymetrix Rat Gene 1.0 ST Array 10920966 - -
          Affymetrix Rat Gene 1.0 ST Array 10920958 - -
          Affymetrix Rat Gene 1.0 ST Array 10920959 - -
          Affymetrix Rat Gene 1.0 ST Array 10920957 - -
          Affymetrix Rat Gene 1.0 ST Array 10920961 - -
          Affymetrix Rat Gene 1.0 ST Array 10920964 - -
          Affymetrix Rat Gene 1.0 ST Array 10920960 - -
          Affymetrix Rat Gene 1.0 ST Array 10920963 - -
          Affymetrix Rat Gene 1.0 ST Array 10920965 - -
          Affymetrix Rat Gene 1.1 ST Array 10920956 - -
          Rat Genome 230 2.0 Array 1372388_at - -
          Rat Genome 230 2.0 Array 1384152_at - -
          Rat Genome 230 2.0 Array 1387000_PM_at - -
          Rat Genome 230 2.0 Array 1384152_PM_at - -
          Rat Genome 230 2.0 Array 1372388_PM_at - -
          Rat Genome 230 2.0 Array 1387000_at - -
          Affymetrix HT Rat Focus Array 1387000_PM_at - -
          Affymetrix HT Rat Focus Array 1387000_at - -
          Affymetrix Rat Clariom S Assay PSR0800028989.rn.1 TC0800003555.rn.1 RTA TC
          Rat Expression Set 230 1387000_at - -
          Rat Expression Set 230 1387000_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920956 - -
          Rat Genome U34 Set rc_AI179027_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20969637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline