Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571440_m1
          See other TGM2 GE Assays ›
          Gene Symbol:
          Tgm2
          Gene Name:
          transglutaminase 2
          Gene Aliases:
          TgaseII
          Chromosome Location:
          Chr.3: 154597165 - 154627257 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571440_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019386.2 NP_062259.2 3 - 4 493 67
          GenBank mRNA AF106325.1 - 2 - 3 444 67
          BC062062.1 - 3 - 4 493 5599202 67

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgm2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          phospholipase C-activating G-protein coupled receptor signaling pathway
          isopeptide cross-linking via N6-(L-isoglutamyl)-L-lysine
          negative regulation of endoplasmic reticulum calcium ion concentration
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          positive regulation of cell adhesion
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          protein homooligomerization
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of mitochondrial calcium ion concentration
          branching involved in salivary gland morphogenesis
          salivary gland cavitation
          protein-glutamine gamma-glutamyltransferase activity
          protein binding
          GTP binding
          enzyme binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P824489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P824489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P824489 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P824489 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1007347 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701615679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88693 - -
          Agilent Rat cDNA Microarray (G4105A) 701615679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851351 - -
          Affymetrix Rat Gene 1.0 ST Array 10851362 - -
          Affymetrix Rat Gene 1.0 ST Array 10851355 - -
          Affymetrix Rat Gene 1.0 ST Array 10851353 - -
          Affymetrix Rat Gene 1.0 ST Array 10851360 - -
          Affymetrix Rat Gene 1.0 ST Array 10851354 - -
          Affymetrix Rat Gene 1.0 ST Array 10851357 - -
          Affymetrix Rat Gene 1.0 ST Array 10851361 - -
          Affymetrix Rat Gene 1.0 ST Array 10851358 - -
          Affymetrix Rat Gene 1.0 ST Array 10851356 - -
          Affymetrix Rat Gene 1.0 ST Array 10851363 - -
          Affymetrix Rat Gene 1.0 ST Array 10851352 - -
          Affymetrix Rat Gene 1.0 ST Array 10851359 - -
          Affymetrix Rat Gene 1.1 ST Array 10851350 - -
          Rat Genome 230 2.0 Array 1387776_PM_at - -
          Rat Genome 230 2.0 Array 1369943_at - -
          Rat Genome 230 2.0 Array 1369943_PM_at - -
          Rat Genome 230 2.0 Array 1387776_at - -
          Affymetrix HT Rat Focus Array 1369943_at - -
          Affymetrix HT Rat Focus Array 1387776_PM_at - -
          Affymetrix HT Rat Focus Array 1369943_PM_at - -
          Affymetrix HT Rat Focus Array 1387776_at - -
          Rat Expression Set 230 1387776_PM_at - -
          Rat Expression Set 230 1369943_PM_at - -
          Rat Expression Set 230 1387776_at - -
          Rat Expression Set 230 1369943_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851350 - -
          Rat Genome U34 Set rc_AI009426_at - -
          Rat Genome U34 Set rc_AI030042_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21308409 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362327 - -
          RatRef-12 Expression BeadChip ILMN_52754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline