Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571479_m1
          See other DLG4 GE Assays ›
          Gene Symbol:
          Dlg4
          Gene Name:
          discs, large homolog 4 (Drosophila)
          Gene Aliases:
          Dlgh4, PSD95, Sap90
          Chromosome Location:
          Chr.10: 56625630 - 56655543 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571479_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019621.1 NP_062567.1 8 - 9 842 91
          XM_006246680.2 XP_006246742.1 10 - 11 1218 91
          XM_006246681.2 XP_006246743.1 10 - 11 1213 91
          XM_006246682.2 XP_006246744.1 10 - 11 1067 91
          XM_006246683.2 XP_006246745.1 10 - 11 975 91
          XM_006246684.2 XP_006246746.1 10 - 11 966 91
          XM_006246685.2 XP_006246747.1 8 - 9 928 91
          XM_006246686.2 XP_006246748.1 8 - 9 988 91
          XM_006246687.2 XP_006246749.1 8 - 9 888 91
          XM_006246688.2 XP_006246750.1 7 - 8 3089 91
          XM_006246689.2 XP_006246751.1 7 - 8 831 91
          XM_006246691.2 XP_006246753.1 7 - 8 822 91
          XM_008767809.1 XP_008766031.1 10 - 11 1319 91
          XM_008767810.1 XP_008766032.1 7 - 8 706 91
          GenBank mRNA M96853.1 - 8 - 9 842 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000945.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dlg4

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of receptor internalization
          protein complex assembly
          positive regulation of cytosolic calcium ion concentration
          synaptic vesicle maturation
          social behavior
          protein localization to synapse
          locomotory exploration behavior
          neuronal ion channel clustering
          establishment of protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          nucleotide phosphorylation
          regulation of long-term neuronal synaptic plasticity
          positive regulation of synaptic transmission
          neuromuscular process controlling balance
          dendritic spine morphogenesis
          vocalization behavior
          AMPA glutamate receptor clustering
          receptor localization to synapse
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          regulation of grooming behavior
          guanylate kinase activity
          receptor binding
          protein binding
          protein C-terminus binding
          protein kinase binding
          protein phosphatase binding
          PDZ domain binding
          beta-1 adrenergic receptor binding
          D1 dopamine receptor binding
          P2Y1 nucleotide receptor binding
          protein complex binding
          acetylcholine receptor binding
          glutamate receptor binding
          ionotropic glutamate receptor binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P238312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238312 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238312 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P238312 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238312 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238314 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700778307 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62602 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44740 - -
          Agilent Rat cDNA Microarray (G4105A) 700778307 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735156 - -
          Affymetrix Rat Gene 1.0 ST Array 10735142 - -
          Affymetrix Rat Gene 1.0 ST Array 10735145 - -
          Affymetrix Rat Gene 1.0 ST Array 10735155 - -
          Affymetrix Rat Gene 1.0 ST Array 10735141 - -
          Affymetrix Rat Gene 1.0 ST Array 10735151 - -
          Affymetrix Rat Gene 1.0 ST Array 10735143 - -
          Affymetrix Rat Gene 1.0 ST Array 10735150 - -
          Affymetrix Rat Gene 1.0 ST Array 10735138 - -
          Affymetrix Rat Gene 1.0 ST Array 10735157 - -
          Affymetrix Rat Gene 1.0 ST Array 10735154 - -
          Affymetrix Rat Gene 1.0 ST Array 10735152 - -
          Affymetrix Rat Gene 1.0 ST Array 10735144 - -
          Affymetrix Rat Gene 1.0 ST Array 10735140 - -
          Affymetrix Rat Gene 1.0 ST Array 10735147 - -
          Affymetrix Rat Gene 1.0 ST Array 10735149 - -
          Affymetrix Rat Gene 1.0 ST Array 10735148 - -
          Affymetrix Rat Gene 1.0 ST Array 10735158 - -
          Affymetrix Rat Gene 1.0 ST Array 10735153 - -
          Affymetrix Rat Gene 1.1 ST Array 10735135 - -
          Rat Genome 230 2.0 Array 1371183_a_at - -
          Rat Genome 230 2.0 Array 1387210_PM_at - -
          Rat Genome 230 2.0 Array 1389901_at - -
          Rat Genome 230 2.0 Array 1371183_PM_a_at - -
          Rat Genome 230 2.0 Array 1389901_PM_at - -
          Rat Genome 230 2.0 Array 1387210_at - -
          Affymetrix HT Rat Focus Array 1389901_PM_at - -
          Affymetrix HT Rat Focus Array 1371183_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371183_a_at - -
          Affymetrix HT Rat Focus Array 1389901_at - -
          Affymetrix HT Rat Focus Array 1387210_at - -
          Affymetrix HT Rat Focus Array 1387210_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000008364.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008355.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008359.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008338.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008360.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008354.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008331.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008361.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008340.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008342.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008368.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008336.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008369.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008366.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008362.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008357.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008358.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008347.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008346.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008351.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008350.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008343.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008353.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008365.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008367.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008370.rn.1 TC1000000945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008333.rn.1 TC1000000945.rn.1 RTA TC
          Rat Expression Set 230 1387210_at - -
          Rat Expression Set 230 1389901_PM_at - -
          Rat Expression Set 230 1389901_at - -
          Rat Expression Set 230 1371183_a_at - -
          Rat Expression Set 230 1387210_PM_at - -
          Rat Expression Set 230 1371183_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735135 - -
          Rat Genome U34 Set M96853_at - -
          Rat Genome U34 Set M96853_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20761529 - -
          ABI Rat Genome Survey Microarray 21009065 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650748 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline