Search
Search
| Gene Symbol: Lgals1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019904.1 | NP_063969.1 | 2 - 3 | 122 | 98 | |
| GenBank mRNA | BC058476.1 | - | 2 - 3 | 152 | 6920799 | 98 |
| FQ217640.1 | - | 2 - 3 | 149 | 98 | ||
| FQ221017.1 | - | 2 - 3 | 148 | 98 | ||
| FQ221731.1 | - | 2 - 3 | 148 | 98 | ||
| FQ221808.1 | - | 2 - 3 | 149 | 98 | ||
| FQ221945.1 | - | 2 - 3 | 149 | 98 | ||
| FQ222079.1 | - | 2 - 3 | 149 | 98 | ||
| FQ222712.1 | - | 2 - 3 | 149 | 98 | ||
| FQ223159.1 | - | 2 - 3 | 148 | 98 | ||
| FQ223277.1 | - | 2 - 3 | 149 | 98 | ||
| FQ223897.1 | - | 2 - 3 | 149 | 98 | ||
| FQ224769.1 | - | 2 - 3 | 149 | 98 | ||
| FQ228416.1 | - | 1 - 2 | 76 | 98 | ||
| FQ228452.1 | - | 2 - 3 | 149 | 98 | ||
| FQ228459.1 | - | 2 - 3 | 148 | 98 | ||
| FQ228622.1 | - | 2 - 3 | 147 | 98 | ||
| FQ230068.1 | - | 2 - 3 | 149 | 98 | ||
| M19036.1 | - | 2 - 3 | 122 | 98 | ||
Set Membership: |
|