Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571632_m1
          See other ADAM2 GE Assays ›
          Gene Symbol:
          Adam2
          Gene Name:
          ADAM metallopeptidase domain 2
          Gene Aliases:
          Ftnb, PH-30
          Chromosome Location:
          Chr.15: 42708536 - 42751281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020077.1 NP_064462.1 18 - 19 2036 54
          GenBank mRNA X99794.1 - 18 - 19 2036 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001854.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cell adhesion
          spermatogenesis
          visual learning
          fertilization
          adult behavior
          metalloendopeptidase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P271511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271511 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P271511 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P271511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784609 - -
          Affymetrix Rat Gene 1.0 ST Array 10784614 - -
          Affymetrix Rat Gene 1.0 ST Array 10784608 - -
          Affymetrix Rat Gene 1.0 ST Array 10784606 - -
          Affymetrix Rat Gene 1.0 ST Array 10784602 - -
          Affymetrix Rat Gene 1.0 ST Array 10784613 - -
          Affymetrix Rat Gene 1.0 ST Array 10784616 - -
          Affymetrix Rat Gene 1.0 ST Array 10784599 - -
          Affymetrix Rat Gene 1.0 ST Array 10784618 - -
          Affymetrix Rat Gene 1.0 ST Array 10784617 - -
          Affymetrix Rat Gene 1.0 ST Array 10784620 - -
          Affymetrix Rat Gene 1.0 ST Array 10784604 - -
          Affymetrix Rat Gene 1.0 ST Array 10784619 - -
          Affymetrix Rat Gene 1.0 ST Array 10784600 - -
          Affymetrix Rat Gene 1.0 ST Array 10784615 - -
          Affymetrix Rat Gene 1.0 ST Array 10784605 - -
          Affymetrix Rat Gene 1.0 ST Array 10784601 - -
          Affymetrix Rat Gene 1.0 ST Array 10784610 - -
          Affymetrix Rat Gene 1.0 ST Array 10784612 - -
          Affymetrix Rat Gene 1.0 ST Array 10784611 - -
          Affymetrix Rat Gene 1.0 ST Array 10784603 - -
          Affymetrix Rat Gene 1.0 ST Array 10784607 - -
          Affymetrix Rat Gene 1.1 ST Array 10784598 - -
          Rat Genome 230 2.0 Array 1369833_at - -
          Rat Genome 230 2.0 Array 1369833_PM_at - -
          Affymetrix HT Rat Focus Array 1369833_PM_at - -
          Affymetrix HT Rat Focus Array 1369833_at - -
          Affymetrix Rat Clariom S Assay PSR1500012434.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012443.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012445.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012447.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012435.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012436.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012442.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012446.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012449.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012451.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012437.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012439.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012431.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012433.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012440.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012448.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012438.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012441.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012453.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012455.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012432.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012452.rn.1 TC1500001854.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012454.rn.1 TC1500001854.rn.1 RTA TC
          Rat Expression Set 230 1369833_at - -
          Rat Expression Set 230 1369833_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784598 - -
          Rat Genome U34 Set X99794cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21669447 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368097 - -
          RatRef-12 Expression BeadChip ILMN_48921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline