Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571731_m1
          See other NOTCH3 GE Assays ›
          Gene Symbol:
          Notch3
          Gene Name:
          notch 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 14138495 - 14189688 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571731_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Notch3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020087.2 NP_064472.2 2 - 3 296 112
          GenBank mRNA AF164486.2 - 2 - 3 296 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002232.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Notch3

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          Notch signaling pathway
          multicellular organism development
          cell differentiation
          forebrain development
          tissue regeneration
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of smooth muscle cell proliferation
          neuron fate commitment
          glomerular capillary formation
          calcium ion binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133410 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133410 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96269 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71441 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900938 - -
          Affymetrix Rat Gene 1.0 ST Array 10900913 - -
          Affymetrix Rat Gene 1.0 ST Array 10894164 - -
          Affymetrix Rat Gene 1.0 ST Array 10900914 - -
          Affymetrix Rat Gene 1.0 ST Array 10900923 - -
          Affymetrix Rat Gene 1.0 ST Array 10900918 - -
          Affymetrix Rat Gene 1.0 ST Array 10900928 - -
          Affymetrix Rat Gene 1.0 ST Array 10900931 - -
          Affymetrix Rat Gene 1.0 ST Array 10900919 - -
          Affymetrix Rat Gene 1.0 ST Array 10900920 - -
          Affymetrix Rat Gene 1.0 ST Array 10900929 - -
          Affymetrix Rat Gene 1.0 ST Array 10900937 - -
          Affymetrix Rat Gene 1.0 ST Array 10900921 - -
          Affymetrix Rat Gene 1.0 ST Array 10900936 - -
          Affymetrix Rat Gene 1.0 ST Array 10900926 - -
          Affymetrix Rat Gene 1.0 ST Array 10900930 - -
          Affymetrix Rat Gene 1.0 ST Array 10900915 - -
          Affymetrix Rat Gene 1.0 ST Array 10900908 - -
          Affymetrix Rat Gene 1.0 ST Array 10900925 - -
          Affymetrix Rat Gene 1.0 ST Array 10900911 - -
          Affymetrix Rat Gene 1.0 ST Array 10900932 - -
          Affymetrix Rat Gene 1.0 ST Array 10900910 - -
          Affymetrix Rat Gene 1.0 ST Array 10900927 - -
          Affymetrix Rat Gene 1.0 ST Array 10900912 - -
          Affymetrix Rat Gene 1.0 ST Array 10900922 - -
          Affymetrix Rat Gene 1.0 ST Array 10900939 - -
          Affymetrix Rat Gene 1.0 ST Array 10900935 - -
          Affymetrix Rat Gene 1.0 ST Array 10900924 - -
          Affymetrix Rat Gene 1.0 ST Array 10900934 - -
          Affymetrix Rat Gene 1.0 ST Array 10900940 - -
          Affymetrix Rat Gene 1.0 ST Array 10900917 - -
          Affymetrix Rat Gene 1.1 ST Array 10900907 - -
          Affymetrix Rat Gene 1.1 ST Array 10894163 - -
          Rat Genome 230 2.0 Array 1369329_at - -
          Rat Genome 230 2.0 Array 1369329_PM_at - -
          Affymetrix HT Rat Focus Array 1369329_at - -
          Affymetrix HT Rat Focus Array 1369329_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700017872.rn.1 TC0700002232.rn.1 RTA TC
          Rat Expression Set 230 1369329_at - -
          Rat Expression Set 230 1369329_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900907 - -
          Rat Genome U34 Set rc_AI102839_at - -
          Rat Genome U34 Set rc_AI102839_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21405916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353245 - -
          RatRef-12 Expression BeadChip ILMN_65743 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline