Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571814_m1
          See other LEPROT GE Assays ›
          Gene Symbol:
          Leprot
          Gene Name:
          leptin receptor overlapping transcript
          Gene Aliases:
          Ob-Rgrp, Obrgrp
          Chromosome Location:
          Chr.5: 120498910 - 120511011 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571814_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Leprot
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020099.2 NP_064484.1 3 - 4 297 81
          GenBank mRNA AF139209.1 - 3 - 4 324 81
          BC062003.1 - 3 - 4 297 5599858 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001077.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Leprot

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          negative regulation of JAK-STAT cascade
          negative regulation of growth hormone receptor signaling pathway
          negative regulation of protein localization to cell surface
          cytokine receptor activity
          receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166000 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155790 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155790 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12067 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P547133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870294 - -
          Affymetrix Rat Gene 1.0 ST Array 10870293 - -
          Affymetrix Rat Gene 1.0 ST Array 10870292 - -
          Affymetrix Rat Gene 1.0 ST Array 10870290 - -
          Affymetrix Rat Gene 1.0 ST Array 10870291 - -
          Affymetrix Rat Gene 1.1 ST Array 10870289 - -
          Rat Genome 230 2.0 Array 1369323_at - -
          Rat Genome 230 2.0 Array 1388725_PM_at - -
          Rat Genome 230 2.0 Array 1388725_at - -
          Rat Genome 230 2.0 Array 1369323_PM_at - -
          Affymetrix HT Rat Focus Array 1369323_PM_at - -
          Affymetrix HT Rat Focus Array 1388725_at - -
          Affymetrix HT Rat Focus Array 1388725_PM_at - -
          Affymetrix HT Rat Focus Array 1369323_at - -
          Affymetrix Rat Clariom S Assay PSR0500006916.rn.1 TC0500001077.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500006917.rn.1 TC0500001077.rn.1 RTA TC
          Rat Expression Set 230 1388725_at - -
          Rat Expression Set 230 1369323_at - -
          Rat Expression Set 230 1369323_PM_at - -
          Rat Expression Set 230 1388725_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870289 - -
          Rat Genome U34 Set rc_AA956453_g_at - -
          Rat Genome U34 Set rc_AA956453_at - -
          Rat Genome U34 Set H32689_at - -
          Rat Genome U34 Set rc_AI231491_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364359 - -
          RatRef-12 Expression BeadChip ILMN_59770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline