Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571880_m1
          See other ADAM17 GE Assays ›
          Gene Symbol:
          Adam17
          Gene Name:
          ADAM metallopeptidase domain 17
          Gene Aliases:
          TACE
          Chromosome Location:
          Chr.6: 43400525 - 43448280 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571880_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020306.2 NP_064702.1 10 - 11 1431 143
          GenBank mRNA AJ012603.1 - 10 - 11 1433 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002209.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam17

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          germinal center formation
          proteolysis
          membrane protein ectodomain proteolysis
          cell adhesion
          epidermal growth factor receptor signaling pathway
          Notch signaling pathway
          Notch receptor processing
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          positive regulation of T cell chemotaxis
          B cell differentiation
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          response to lipopolysaccharide
          negative regulation of interleukin-8 production
          positive regulation of chemokine production
          regulation of mast cell apoptotic process
          T cell differentiation in thymus
          cell adhesion mediated by integrin
          wound healing, spreading of epidermal cells
          epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
          response to drug
          negative regulation of apoptotic process
          positive regulation of epidermal growth factor-activated receptor activity
          spleen development
          cell motility
          PMA-inducible membrane protein ectodomain proteolysis
          positive regulation of cellular component movement
          response to high density lipoprotein particle
          metalloendopeptidase activity
          Notch binding
          interleukin-6 receptor binding
          integrin binding
          metallopeptidase activity
          zinc ion binding
          SH3 domain binding
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P517650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517650 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P517650 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4757 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79376 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889348 - -
          Affymetrix Rat Gene 1.0 ST Array 10889346 - -
          Affymetrix Rat Gene 1.0 ST Array 10889357 - -
          Affymetrix Rat Gene 1.0 ST Array 10889347 - -
          Affymetrix Rat Gene 1.0 ST Array 10889358 - -
          Affymetrix Rat Gene 1.0 ST Array 10889343 - -
          Affymetrix Rat Gene 1.0 ST Array 10889355 - -
          Affymetrix Rat Gene 1.0 ST Array 10889356 - -
          Affymetrix Rat Gene 1.0 ST Array 10889345 - -
          Affymetrix Rat Gene 1.0 ST Array 10889350 - -
          Affymetrix Rat Gene 1.0 ST Array 10889354 - -
          Affymetrix Rat Gene 1.0 ST Array 10889340 - -
          Affymetrix Rat Gene 1.0 ST Array 10889344 - -
          Affymetrix Rat Gene 1.0 ST Array 10889342 - -
          Affymetrix Rat Gene 1.0 ST Array 10889353 - -
          Affymetrix Rat Gene 1.0 ST Array 10889352 - -
          Affymetrix Rat Gene 1.0 ST Array 10889351 - -
          Affymetrix Rat Gene 1.0 ST Array 10889341 - -
          Affymetrix Rat Gene 1.0 ST Array 10889349 - -
          Affymetrix Rat Gene 1.0 ST Array 10889359 - -
          Affymetrix Rat Gene 1.1 ST Array 10889339 - -
          Rat Genome 230 2.0 Array 1394496_PM_at - -
          Rat Genome 230 2.0 Array 1367922_PM_at - -
          Rat Genome 230 2.0 Array 1394496_at - -
          Rat Genome 230 2.0 Array 1367922_at - -
          Affymetrix HT Rat Focus Array 1367922_PM_at - -
          Affymetrix HT Rat Focus Array 1367922_at - -
          Affymetrix Rat Clariom S Assay PSR0600015995.rn.1 TC0600002209.rn.1 RTA TC
          Rat Expression Set 230 1367922_at - -
          Rat Expression Set 230 1367922_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889339 - -
          Rat Genome U34 Set AJ012603cds_at - -
          Rat Genome U34 Set AJ012603UTR#1_g_at - -
          Rat Genome U34 Set AJ012603UTR#1_at - -
          Rat Genome U34 Set AJ012603UTR#1_g_at - -
          Rat Genome U34 Set AJ012603cds_at - -
          Rat Genome U34 Set AJ012603UTR#1_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21920715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360815 - -
          RatRef-12 Expression BeadChip ILMN_53540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0