Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572047_m1
          See other PTGES GE Assays ›
          Gene Symbol:
          Ptges
          Gene Name:
          prostaglandin E synthase
          Gene Aliases:
          Pges
          Chromosome Location:
          Chr.3: 9727408 - 9738752 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572047_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptges
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021583.3 NP_067594.1 2 - 3 254 66
          GenBank mRNA AB035145.1 - 2 - 3 214 66
          AB041998.1 - 2 - 3 243 66
          AB048730.1 - 2 - 3 256 66
          AF280967.1 - 2 - 3 253 66
          BC088101.1 - 2 - 3 212 66
          BC105858.1 - 2 - 3 240 7456259 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002838.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptges

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          acute inflammatory response
          chronic inflammatory response
          prostaglandin metabolic process
          negative regulation of cell proliferation
          response to organic cyclic compound
          response to lipopolysaccharide
          response to retinoic acid
          response to cytokine
          response to calcium ion
          glutathione binding
          prostaglandin-E synthase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P163784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953404 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12079 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12079 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701301785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14937 - -
          Agilent Rat cDNA Microarray (G4105A) 701301785 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844226 - -
          Affymetrix Rat Gene 1.0 ST Array 10844225 - -
          Affymetrix Rat Gene 1.0 ST Array 10844228 - -
          Affymetrix Rat Gene 1.0 ST Array 10844224 - -
          Affymetrix Rat Gene 1.0 ST Array 10844227 - -
          Affymetrix Rat Gene 1.1 ST Array 10844223 - -
          Rat Genome 230 2.0 Array 1368014_at - -
          Rat Genome 230 2.0 Array 1368015_PM_at - -
          Rat Genome 230 2.0 Array 1368014_PM_at - -
          Rat Genome 230 2.0 Array 1368015_at - -
          Affymetrix HT Rat Focus Array 1368015_at - -
          Affymetrix HT Rat Focus Array 1368014_at - -
          Affymetrix HT Rat Focus Array 1368015_PM_at - -
          Affymetrix HT Rat Focus Array 1368014_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300021696.rn.1 TC0300002838.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021697.rn.1 TC0300002838.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021694.rn.1 TC0300002838.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021695.rn.1 TC0300002838.rn.1 RTA TC
          Rat Expression Set 230 1368015_at - -
          Rat Expression Set 230 1368015_PM_at - -
          Rat Expression Set 230 1368014_at - -
          Rat Expression Set 230 1368014_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844223 - -
          Rat Genome U34 Set rc_AA944447_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22382100 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353392 - -
          RatRef-12 Expression BeadChip ILMN_49901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline