Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572130_m1
          See other NTRK1 GE Assays ›
          Gene Symbol:
          Ntrk1
          Gene Name:
          neurotrophic tyrosine kinase, receptor, type 1
          Gene Aliases:
          Trk
          Chromosome Location:
          Chr.2: 187143562 - 187160373 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572130_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ntrk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021589.1 NP_067600.1 6 - 7 783 65
          XM_006232746.2 XP_006232808.1 6 - 7 774 65
          GenBank mRNA M85214.1 - 6 - 7 783 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004013.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ntrk1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          nervous system development
          axon guidance
          aging
          learning or memory
          circadian rhythm
          negative regulation of cell proliferation
          response to radiation
          programmed cell death involved in cell development
          positive regulation of neuron projection development
          response to activity
          peptidyl-tyrosine phosphorylation
          sensory perception of pain
          olfactory nerve development
          B cell differentiation
          response to nutrient levels
          response to nicotine
          nerve growth factor signaling pathway
          mechanoreceptor differentiation
          response to drug
          positive regulation of programmed cell death
          negative regulation of neuron apoptotic process
          response to ethanol
          positive regulation of angiogenesis
          positive regulation of Ras protein signal transduction
          protein autophosphorylation
          neurotrophin TRK receptor signaling pathway
          sympathetic nervous system development
          response to axon injury
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of NF-kappaB transcription factor activity
          response to hydrostatic pressure
          response to electrical stimulus
          positive regulation of synaptic transmission, glutamatergic
          Sertoli cell development
          axonogenesis involved in innervation
          positive regulation of ERK1 and ERK2 cascade
          cellular response to nicotine
          cellular response to growth factor stimulus
          negative regulation of neuron death
          cellular response to nerve growth factor stimulus
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          neurotrophin p75 receptor binding
          protein binding
          ATP binding
          nerve growth factor receptor activity
          kinase binding
          protein homodimerization activity
          ephrin receptor binding
          nerve growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115417 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P430547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117656 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115417 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P430547 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P430547 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115417 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115417 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P430547 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P780039 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P430547 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6435 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824203 - -
          Affymetrix Rat Gene 1.0 ST Array 10824198 - -
          Affymetrix Rat Gene 1.0 ST Array 10824201 - -
          Affymetrix Rat Gene 1.0 ST Array 10824206 - -
          Affymetrix Rat Gene 1.0 ST Array 10824196 - -
          Affymetrix Rat Gene 1.0 ST Array 10824192 - -
          Affymetrix Rat Gene 1.0 ST Array 10824199 - -
          Affymetrix Rat Gene 1.0 ST Array 10824190 - -
          Affymetrix Rat Gene 1.0 ST Array 10824204 - -
          Affymetrix Rat Gene 1.0 ST Array 10824195 - -
          Affymetrix Rat Gene 1.0 ST Array 10824197 - -
          Affymetrix Rat Gene 1.0 ST Array 10824202 - -
          Affymetrix Rat Gene 1.0 ST Array 10824189 - -
          Affymetrix Rat Gene 1.0 ST Array 10824193 - -
          Affymetrix Rat Gene 1.0 ST Array 10824194 - -
          Affymetrix Rat Gene 1.0 ST Array 10824200 - -
          Affymetrix Rat Gene 1.0 ST Array 10824205 - -
          Affymetrix Rat Gene 1.0 ST Array 10824191 - -
          Affymetrix Rat Gene 1.1 ST Array 10824188 - -
          Rat Genome 230 2.0 Array 1369000_PM_at - -
          Rat Genome 230 2.0 Array 1375615_PM_at - -
          Rat Genome 230 2.0 Array 1369000_at - -
          Rat Genome 230 2.0 Array 1375615_at - -
          Affymetrix HT Rat Focus Array 1375615_PM_at - -
          Affymetrix HT Rat Focus Array 1375615_at - -
          Affymetrix HT Rat Focus Array 1369000_PM_at - -
          Affymetrix HT Rat Focus Array 1369000_at - -
          Affymetrix Rat Clariom S Assay PSR0200027043.rn.1 TC0200004013.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027044.rn.1 TC0200004013.rn.1 RTA TC
          Rat Expression Set 230 1375615_at - -
          Rat Expression Set 230 1369000_at - -
          Rat Expression Set 230 1375615_PM_at - -
          Rat Expression Set 230 1369000_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824188 - -
          Rat Genome U34 Set M85214_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22103001 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370831 - -
          RatRef-12 Expression BeadChip ILMN_58089 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline