Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572135_m1
          See other AIPL1 GE Assays ›
          Gene Symbol:
          Aipl1
          Gene Name:
          aryl hydrocarbon receptor-interacting protein-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 58579129 - 58631194 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572135_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aipl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021590.1 NP_067601.1 2 - 3 337 62
          XM_006246798.2 XP_006246860.1 2 - 3 369 62
          GenBank mRNA AF180340.1 - 2 - 3 337 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003168.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aipl1

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          protein folding
          phototransduction, visible light
          biological_process
          protein farnesylation
          regulation of cGMP metabolic process
          negative regulation of apoptotic process
          farnesylated protein binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063249 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744634 - -
          Affymetrix Rat Gene 1.0 ST Array 10744637 - -
          Affymetrix Rat Gene 1.0 ST Array 10744638 - -
          Affymetrix Rat Gene 1.0 ST Array 10744633 - -
          Affymetrix Rat Gene 1.0 ST Array 10744640 - -
          Affymetrix Rat Gene 1.0 ST Array 10744635 - -
          Affymetrix Rat Gene 1.0 ST Array 10744639 - -
          Affymetrix Rat Gene 1.1 ST Array 10744632 - -
          Rat Genome 230 2.0 Array 1387166_PM_at - -
          Rat Genome 230 2.0 Array 1387166_at - -
          Affymetrix HT Rat Focus Array 1387166_PM_at - -
          Affymetrix HT Rat Focus Array 1387166_at - -
          Affymetrix Rat Clariom S Assay PSR1000028519.rn.1 TC1000003168.rn.1 RTA TC
          Rat Expression Set 230 1387166_PM_at - -
          Rat Expression Set 230 1387166_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744632 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22146264 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365560 - -
          RatRef-12 Expression BeadChip ILMN_48515 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline