Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572154_m1
          See other SLC9A3R1 GE Assays ›
          Gene Symbol:
          Slc9a3r1
          Gene Name:
          solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 103713045 - 103730145 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572154_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc9a3r1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021594.1 NP_067605.1 1 - 2 508 93
          GenBank mRNA AF154336.1 - 1 - 2 508 93
          FQ213792.1 - 1 - 2 625 93
          FQ228926.1 - 1 - 2 606 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001917.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc9a3r1

          Gene Ontology Categories:

          Function(s) Process(es)

          renal sodium ion transport
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of sound
          negative regulation of cell proliferation
          negative regulation of platelet-derived growth factor receptor signaling pathway
          negative regulation of sodium ion transport
          negative regulation of phosphatidylinositol 3-kinase signaling
          Wnt signaling pathway
          cAMP-mediated signaling
          microvillus assembly
          actin cytoskeleton organization
          negative regulation of cell migration
          cellular phosphate ion homeostasis
          negative regulation of sodium:proton antiporter activity
          bile acid secretion
          cellular protein localization
          glutathione transport
          regulation of excretion
          regulation of protein kinase activity
          negative regulation of protein kinase B signaling
          auditory receptor cell stereocilium organization
          phospholipase C-activating dopamine receptor signaling pathway
          renal absorption
          negative regulation of ERK1 and ERK2 cascade
          renal phosphate ion absorption
          negative regulation of cell motility
          positive regulation of intrinsic apoptotic signaling pathway
          receptor binding
          protein binding
          beta-catenin binding
          chloride channel regulator activity
          phosphatase binding
          protein domain specific binding
          PDZ domain binding
          beta-2 adrenergic receptor binding
          protein complex binding
          protein complex scaffold
          protein self-association
          dopamine receptor binding
          growth factor receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P136603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P136603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P136603 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P136603 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P136603 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700501829 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23259 - -
          Agilent Rat cDNA Microarray (G4105A) 700330190 - -
          Agilent Rat cDNA Microarray (G4105A) 700501829 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739496 - -
          Affymetrix Rat Gene 1.0 ST Array 10739493 - -
          Affymetrix Rat Gene 1.0 ST Array 10739494 - -
          Affymetrix Rat Gene 1.0 ST Array 10739491 - -
          Affymetrix Rat Gene 1.0 ST Array 10739495 - -
          Affymetrix Rat Gene 1.0 ST Array 10739492 - -
          Affymetrix Rat Gene 1.1 ST Array 10739490 - -
          Rat Genome 230 2.0 Array 1370032_at - -
          Rat Genome 230 2.0 Array 1387793_at - -
          Rat Genome 230 2.0 Array 1370032_PM_at - -
          Rat Genome 230 2.0 Array 1387793_PM_at - -
          Affymetrix HT Rat Focus Array 1387793_PM_at - -
          Affymetrix HT Rat Focus Array 1370032_PM_at - -
          Affymetrix HT Rat Focus Array 1387793_at - -
          Affymetrix HT Rat Focus Array 1370032_at - -
          Affymetrix Rat Clariom S Assay PSR1000017222.rn.1 TC1000001917.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000017221.rn.1 TC1000001917.rn.1 RTA TC
          Rat Expression Set 230 1370032_PM_at - -
          Rat Expression Set 230 1370032_at - -
          Rat Expression Set 230 1387793_PM_at - -
          Rat Expression Set 230 1387793_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739490 - -
          Rat Genome U34 Set rc_AI007803_at - -
          Rat Genome U34 Set rc_AI233724_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21561968 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364745 - -
          RatRef-12 Expression BeadChip ILMN_51318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline