Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572211_m1
          See other PHLPP1 GE Assays ›
          Gene Symbol:
          Phlpp1
          Gene Name:
          PH domain and leucine rich repeat protein phosphatase 1
          Gene Aliases:
          Phlpp, Plekhe1, Scop
          Chromosome Location:
          Chr.13: 26145989 - 26394505 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phlpp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021657.1 NP_067689.1 9 - 10 2948 95
          GenBank mRNA AB023624.1 - 9 - 10 2948 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000077.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phlpp1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          protein dephosphorylation
          apoptotic process
          circadian rhythm
          metabolic process
          entrainment of circadian clock
          hippocampus development
          negative regulation of protein kinase B signaling
          negative regulation of protein kinase C signaling
          catalytic activity
          protein serine/threonine phosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P661018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42367 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88641 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P661018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P661018 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P661018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P661018 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77841 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94734 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95832 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18131 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87708 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17043 - -
          Agilent Rat cDNA Microarray (G4105A) 700695107 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763304 - -
          Affymetrix Rat Gene 1.0 ST Array 10763309 - -
          Affymetrix Rat Gene 1.0 ST Array 10763300 - -
          Affymetrix Rat Gene 1.0 ST Array 10763312 - -
          Affymetrix Rat Gene 1.0 ST Array 10763301 - -
          Affymetrix Rat Gene 1.0 ST Array 10763310 - -
          Affymetrix Rat Gene 1.0 ST Array 10763315 - -
          Affymetrix Rat Gene 1.0 ST Array 10763308 - -
          Affymetrix Rat Gene 1.0 ST Array 10763313 - -
          Affymetrix Rat Gene 1.0 ST Array 10763311 - -
          Affymetrix Rat Gene 1.0 ST Array 10763314 - -
          Affymetrix Rat Gene 1.0 ST Array 10763306 - -
          Affymetrix Rat Gene 1.0 ST Array 10763303 - -
          Affymetrix Rat Gene 1.0 ST Array 10763307 - -
          Affymetrix Rat Gene 1.0 ST Array 10763302 - -
          Affymetrix Rat Gene 1.0 ST Array 10763305 - -
          Affymetrix Rat Gene 1.1 ST Array 10763299 - -
          Rat Genome 230 2.0 Array 1368262_PM_at - -
          Rat Genome 230 2.0 Array 1368262_at - -
          Affymetrix HT Rat Focus Array 1368262_PM_at - -
          Affymetrix HT Rat Focus Array 1368262_at - -
          Affymetrix Rat Clariom S Assay PSR1300000375.rn.1 TC1300000077.rn.1 RTA TC
          Rat Expression Set 230 1368262_at - -
          Rat Expression Set 230 1368262_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763299 - -
          Rat Genome U34 Set rc_AA819723_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21172268 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362245 - -
          RatRef-12 Expression BeadChip ILMN_57175 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline