Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572484_m1
          See other SMAD5 GE Assays ›
          Gene Symbol:
          Smad5
          Gene Name:
          SMAD family member 5
          Gene Aliases:
          Madh5
          Chromosome Location:
          Chr.17: 8307451 - 8336996 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572484_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021692.1 NP_067724.1 2 - 3 734 90
          XM_006253586.2 XP_006253648.1 5 - 6 1199 90
          XM_006253587.2 XP_006253649.1 4 - 5 1117 90
          XM_006253588.2 XP_006253650.1 5 - 6 1441 90
          XM_008771446.1 XP_008769668.1 4 - 5 2777 90
          XM_008771447.1 XP_008769669.1 5 - 6 1959 90
          XM_008771448.1 XP_008769670.1 5 - 6 2134 90
          GenBank mRNA AB010955.1 - 2 - 3 734 90
          BC070905.1 - 5 - 6 1129 90
          BC161849.1 - 3 - 4 921 9099068 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001281.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad5

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          ureteric bud development
          Mullerian duct regression
          osteoblast fate commitment
          transcription, DNA-templated
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          germ cell development
          embryonic pattern specification
          erythrocyte differentiation
          regulation of ossification
          BMP signaling pathway
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cartilage development
          cardiac muscle contraction
          bone development
          cellular response to organic cyclic compound
          cellular response to BMP stimulus
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          ubiquitin protein ligase binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164787 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14871 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797130 - -
          Affymetrix Rat Gene 1.0 ST Array 10797136 - -
          Affymetrix Rat Gene 1.0 ST Array 10797135 - -
          Affymetrix Rat Gene 1.0 ST Array 10797133 - -
          Affymetrix Rat Gene 1.0 ST Array 10797132 - -
          Affymetrix Rat Gene 1.0 ST Array 10797131 - -
          Affymetrix Rat Gene 1.0 ST Array 10797128 - -
          Affymetrix Rat Gene 1.0 ST Array 10797129 - -
          Affymetrix Rat Gene 1.0 ST Array 10797134 - -
          Affymetrix Rat Gene 1.1 ST Array 10797127 - -
          Rat Genome 230 2.0 Array 1390373_PM_at - -
          Rat Genome 230 2.0 Array 1369276_at - -
          Rat Genome 230 2.0 Array 1390373_at - -
          Rat Genome 230 2.0 Array 1369276_PM_at - -
          Affymetrix HT Rat Focus Array 1369276_at - -
          Affymetrix HT Rat Focus Array 1369276_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700008774.rn.1 TC1700001281.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008776.rn.1 TC1700001281.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008775.rn.1 TC1700001281.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008777.rn.1 TC1700001281.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008773.rn.1 TC1700001281.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008778.rn.1 TC1700001281.rn.1 RTA TC
          Rat Expression Set 230 1369276_PM_at - -
          Rat Expression Set 230 1390373_at - -
          Rat Expression Set 230 1390373_PM_at - -
          Rat Expression Set 230 1369276_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797127 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21412829 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350028 - -
          RatRef-12 Expression BeadChip ILMN_68848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline