Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572531_s1
          See other SYNPO GE Assays ›
          Gene Symbol:
          Synpo
          Gene Name:
          synaptopodin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 55870720 - 55891710 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572531_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Synpo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021695.2 NP_067727.2 2 1928 88
          GenBank mRNA AB013130.1 - 1 1454 88
          AM980944.1 - 1398 88
          DY313339.1 - 1 546 88
          EV779156.1 - 1 307 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Synpo

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of actin filament bundle assembly
          regulation of stress fiber assembly
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P553885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157637 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P553885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P553885 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P553885 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P553885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36787 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10804578 - -
          Affymetrix Rat Gene 1.0 ST Array 10804577 - -
          Affymetrix Rat Gene 1.0 ST Array 10804575 - -
          Affymetrix Rat Gene 1.0 ST Array 10804579 - -
          Affymetrix Rat Gene 1.0 ST Array 10804580 - -
          Affymetrix Rat Gene 1.1 ST Array 10804567 - -
          Rat Genome 230 2.0 Array 1387501_PM_at - -
          Rat Genome 230 2.0 Array 1387501_at - -
          Affymetrix HT Rat Focus Array 1387501_at - -
          Affymetrix HT Rat Focus Array 1387501_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800010552.rn.1 TC1800001497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800010553.rn.1 TC1800001497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800010558.rn.1 TC1800001497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800010551.rn.1 TC1800001497.rn.1 RTA TC
          Rat Expression Set 230 1387501_at - -
          Rat Expression Set 230 1387501_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10804567 - -
          Rat Genome U34 Set AB013130_at - -
          Rat Genome U34 Set AB013130_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20968338 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368169 - -
          RatRef-12 Expression BeadChip ILMN_59961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline