Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572611_m1
          See other AKAP14 GE Assays ›
          Gene Symbol:
          Akap14
          Gene Name:
          A kinase (PRKA) anchor protein 14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 123773430 - 123788898 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akap14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021703.1 NP_067735.1 4 - 5 1601 91
          GenBank mRNA AJ002474.1 - 4 - 5 1601 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002359.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akap14

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          protein kinase A binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P117744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P117744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P117744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P117744 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P117744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931999 - -
          Affymetrix Rat Gene 1.0 ST Array 10931997 - -
          Affymetrix Rat Gene 1.0 ST Array 10931996 - -
          Affymetrix Rat Gene 1.0 ST Array 10931998 - -
          Affymetrix Rat Gene 1.0 ST Array 10932001 - -
          Affymetrix Rat Gene 1.0 ST Array 10932000 - -
          Affymetrix Rat Gene 1.1 ST Array 10931995 - -
          Rat Genome 230 2.0 Array 1390229_at - -
          Rat Genome 230 2.0 Array 1369470_at - -
          Rat Genome 230 2.0 Array 1369470_PM_at - -
          Rat Genome 230 2.0 Array 1390229_PM_at - -
          Affymetrix HT Rat Focus Array 1369470_PM_at - -
          Affymetrix HT Rat Focus Array 1369470_at - -
          Affymetrix Rat Clariom S Assay PSR0X00014779.rn.1 TC0X00002359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00014778.rn.1 TC0X00002359.rn.1 RTA TC
          Rat Expression Set 230 1369470_at - -
          Rat Expression Set 230 1369470_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931995 - -
          Rat Genome U34 Set AJ002474cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20743003 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363467 - -
          RatRef-12 Expression BeadChip ILMN_69396 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline