Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572627_m1
          See other CAMK2B GE Assays ›
          Gene Symbol:
          Camk2b
          Gene Name:
          calcium/calmodulin-dependent protein kinase II beta
          Gene Aliases:
          Ck2b
          Chromosome Location:
          Chr.14: 86208876 - 86297751 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572627_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camk2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042354.1 NP_001035813.1 6 - 7 497 88
          NM_001042356.1 NP_001035815.1 6 - 7 497 88
          NM_021739.2 NP_068507.2 6 - 7 497 88
          XM_006251170.2 XP_006251232.1 6 - 7 596 88
          XM_006251171.2 XP_006251233.1 6 - 7 596 88
          XM_006251172.2 XP_006251234.1 6 - 7 596 88
          XM_006251174.2 XP_006251236.1 6 - 7 596 88
          XM_006251176.2 XP_006251238.1 6 - 7 577 88
          XM_006251177.2 XP_006251239.1 6 - 7 576 88
          GenBank mRNA CB616255.1 - 6 - 7 497 88
          M16112.1 - 6 - 7 477 88
          X83375.1 - 6 - 7 459 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002188.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camk2b

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          inhibitory G-protein coupled receptor phosphorylation
          protein phosphorylation
          calcium ion transport
          positive regulation of neuron projection development
          peptidyl-serine phosphorylation
          regulation of synaptic transmission, cholinergic
          positive regulation of phospholipase A2 activity
          response to cadmium ion
          protein autophosphorylation
          regulation of long-term neuronal synaptic plasticity
          cell projection morphogenesis
          neuromuscular process controlling balance
          long-term synaptic potentiation
          activation of meiosis involved in egg activation
          positive regulation of dendritic spine morphogenesis
          positive regulation of synapse maturation
          positive regulation of apoptotic signaling pathway
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          protein binding
          calmodulin binding
          ATP binding
          protein homodimerization activity
          phospholipase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092342 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P452145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77114 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452145 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P452145 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P561102 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42814 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42826 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55540 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778294 - -
          Affymetrix Rat Gene 1.0 ST Array 10778280 - -
          Affymetrix Rat Gene 1.0 ST Array 10778287 - -
          Affymetrix Rat Gene 1.0 ST Array 10778290 - -
          Affymetrix Rat Gene 1.0 ST Array 10778278 - -
          Affymetrix Rat Gene 1.0 ST Array 10778283 - -
          Affymetrix Rat Gene 1.0 ST Array 10778273 - -
          Affymetrix Rat Gene 1.0 ST Array 10778275 - -
          Affymetrix Rat Gene 1.0 ST Array 10778281 - -
          Affymetrix Rat Gene 1.0 ST Array 10778274 - -
          Affymetrix Rat Gene 1.0 ST Array 10778284 - -
          Affymetrix Rat Gene 1.0 ST Array 10778276 - -
          Affymetrix Rat Gene 1.0 ST Array 10778293 - -
          Affymetrix Rat Gene 1.0 ST Array 10778288 - -
          Affymetrix Rat Gene 1.0 ST Array 10778277 - -
          Affymetrix Rat Gene 1.0 ST Array 10778271 - -
          Affymetrix Rat Gene 1.0 ST Array 10778286 - -
          Affymetrix Rat Gene 1.0 ST Array 10778291 - -
          Affymetrix Rat Gene 1.0 ST Array 10778279 - -
          Affymetrix Rat Gene 1.0 ST Array 10778292 - -
          Affymetrix Rat Gene 1.0 ST Array 10778285 - -
          Affymetrix Rat Gene 1.0 ST Array 10778269 - -
          Affymetrix Rat Gene 1.0 ST Array 10778282 - -
          Affymetrix Rat Gene 1.0 ST Array 10778289 - -
          Affymetrix Rat Gene 1.0 ST Array 10778272 - -
          Affymetrix Rat Gene 1.0 ST Array 10778270 - -
          Affymetrix Rat Gene 1.1 ST Array 10778268 - -
          Rat Genome 230 2.0 Array 1375575_at - -
          Rat Genome 230 2.0 Array 1398251_a_at - -
          Rat Genome 230 2.0 Array 1398251_PM_a_at - -
          Rat Genome 230 2.0 Array 1375575_PM_at - -
          Affymetrix HT Rat Focus Array 1398251_PM_a_at - -
          Affymetrix HT Rat Focus Array 1375575_PM_at - -
          Affymetrix HT Rat Focus Array 1398251_a_at - -
          Affymetrix HT Rat Focus Array 1375575_at - -
          Affymetrix Rat Clariom S Assay PSR1400016268.rn.1 TC1400002188.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400016269.rn.1 TC1400002188.rn.1 RTA TC
          Rat Expression Set 230 1398251_a_at - -
          Rat Expression Set 230 1398251_PM_a_at - -
          Rat Expression Set 230 1375575_PM_at - -
          Rat Expression Set 230 1375575_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778268 - -
          Rat Genome U34 Set M16112_g_at - -
          Rat Genome U34 Set M16112_at - -
          Rat Genome U34 Set rc_AI101481_at - -
          Rat Genome U34 Set M16112_at - -
          Rat Genome U34 Set M16112_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21988075 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367246 - -
          RatRef-12 Expression BeadChip ILMN_59957 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline