Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572720_m1
          See other PROM1 GE Assays ›
          Gene Symbol:
          Prom1
          Gene Name:
          prominin 1
          Gene Aliases:
          CD133, Prom
          Chromosome Location:
          Chr.14: 71532547 - 71637400 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prom1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110137.1 NP_001103607.1 18 - 19 2258 57
          NM_021751.2 NP_068519.2 19 - 20 2325 57
          XM_008770211.1 XP_008768433.1 20 - 21 2393 57
          XM_008770212.1 XP_008768434.1 20 - 21 2689 57
          XM_008770213.1 XP_008768435.1 20 - 21 2585 57
          XM_008770214.1 XP_008768436.1 20 - 21 2393 57
          XM_008770215.1 XP_008768437.1 19 - 20 2661 57
          XM_008770216.1 XP_008768438.1 20 - 21 2345 57
          XM_008770217.1 XP_008768439.1 20 - 21 2393 57
          XM_008770218.1 XP_008768440.1 19 - 20 2366 57
          GenBank mRNA AF263368.1 - 18 - 19 2291 57
          AF386758.1 - 18 - 19 2204 57
          AY262731.1 - 18 - 19 2204 57
          DQ871262.1 - 12 - 13 1158 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000718.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prom1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          retina layer formation
          photoreceptor cell maintenance
          retina morphogenesis in camera-type eye
          camera-type eye photoreceptor cell differentiation
          glomerular visceral epithelial cell differentiation
          glomerular parietal epithelial cell differentiation
          positive regulation of nephron tubule epithelial cell differentiation
          molecular_function
          actinin binding
          cadherin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020244 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93057 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82180 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073904 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12115 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P448717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72559 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86984 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88850 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40561 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773126 - -
          Affymetrix Rat Gene 1.0 ST Array 10773139 - -
          Affymetrix Rat Gene 1.0 ST Array 10773120 - -
          Affymetrix Rat Gene 1.0 ST Array 10773118 - -
          Affymetrix Rat Gene 1.0 ST Array 10773133 - -
          Affymetrix Rat Gene 1.0 ST Array 10773134 - -
          Affymetrix Rat Gene 1.0 ST Array 10773136 - -
          Affymetrix Rat Gene 1.0 ST Array 10773117 - -
          Affymetrix Rat Gene 1.0 ST Array 10773129 - -
          Affymetrix Rat Gene 1.0 ST Array 10773137 - -
          Affymetrix Rat Gene 1.0 ST Array 10773144 - -
          Affymetrix Rat Gene 1.0 ST Array 10773130 - -
          Affymetrix Rat Gene 1.0 ST Array 10773127 - -
          Affymetrix Rat Gene 1.0 ST Array 10773116 - -
          Affymetrix Rat Gene 1.0 ST Array 10773119 - -
          Affymetrix Rat Gene 1.0 ST Array 10773132 - -
          Affymetrix Rat Gene 1.0 ST Array 10773128 - -
          Affymetrix Rat Gene 1.0 ST Array 10773141 - -
          Affymetrix Rat Gene 1.0 ST Array 10773138 - -
          Affymetrix Rat Gene 1.0 ST Array 10773145 - -
          Affymetrix Rat Gene 1.0 ST Array 10773140 - -
          Affymetrix Rat Gene 1.0 ST Array 10773123 - -
          Affymetrix Rat Gene 1.0 ST Array 10773124 - -
          Affymetrix Rat Gene 1.0 ST Array 10773125 - -
          Affymetrix Rat Gene 1.0 ST Array 10773135 - -
          Affymetrix Rat Gene 1.0 ST Array 10773122 - -
          Affymetrix Rat Gene 1.0 ST Array 10773143 - -
          Affymetrix Rat Gene 1.0 ST Array 10773131 - -
          Affymetrix Rat Gene 1.1 ST Array 10773115 - -
          Rat Genome 230 2.0 Array 1368008_PM_at - -
          Rat Genome 230 2.0 Array 1368008_at - -
          Affymetrix HT Rat Focus Array 1368008_PM_at - -
          Affymetrix HT Rat Focus Array 1368008_at - -
          Affymetrix Rat Clariom S Assay PSR1400004930.rn.1 TC1400000718.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004931.rn.1 TC1400000718.rn.1 RTA TC
          Rat Expression Set 230 1368008_at - -
          Rat Expression Set 230 1368008_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773115 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21957931 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364705 - -
          RatRef-12 Expression BeadChip ILMN_53481 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0