Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572869_m1
          See other RBCK1 GE Assays ›
          Gene Symbol:
          Rbck1
          Gene Name:
          RanBP-type and C3HC4-type zinc finger containing 1
          Gene Aliases:
          HOIL-1, Pkcbpb15
          Chromosome Location:
          Chr.3: 147793014 - 147809942 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572869_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbck1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021764.1 NP_068532.2 8 - 9 1322 69
          XM_006235252.2 XP_006235314.1 9 - 10 1247 69
          GenBank mRNA AB011369.1 - 7 - 8 1161 69
          BC061723.1 - 8 - 9 1256 5598480 69
          U48248.1 - 8 - 9 1322 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004467.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbck1

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of necroptotic process
          protein linear polyubiquitination
          positive regulation of extrinsic apoptotic signaling pathway
          double-stranded DNA binding
          ubiquitin-protein transferase activity
          protein kinase C binding
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97416 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130588 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130588 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P131897 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700248864 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700247652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59396 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77938 - -
          Agilent Rat cDNA Microarray (G4105A) 700248864 - -
          Agilent Rat cDNA Microarray (G4105A) 700247652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850766 - -
          Affymetrix Rat Gene 1.0 ST Array 10850773 - -
          Affymetrix Rat Gene 1.0 ST Array 10850771 - -
          Affymetrix Rat Gene 1.0 ST Array 10850772 - -
          Affymetrix Rat Gene 1.0 ST Array 10850765 - -
          Affymetrix Rat Gene 1.0 ST Array 10850764 - -
          Affymetrix Rat Gene 1.0 ST Array 10850774 - -
          Affymetrix Rat Gene 1.0 ST Array 10850768 - -
          Affymetrix Rat Gene 1.0 ST Array 10850767 - -
          Affymetrix Rat Gene 1.0 ST Array 10850763 - -
          Affymetrix Rat Gene 1.0 ST Array 10850769 - -
          Affymetrix Rat Gene 1.0 ST Array 10850770 - -
          Affymetrix Rat Gene 1.1 ST Array 10850762 - -
          Rat Genome 230 2.0 Array 1386966_PM_a_at - -
          Rat Genome 230 2.0 Array 1386966_a_at - -
          Affymetrix HT Rat Focus Array 1386966_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386966_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300034306.rn.1 TC0300004467.rn.1 RTA TC
          Rat Expression Set 230 1386966_a_at - -
          Rat Expression Set 230 1386966_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10850762 - -
          Rat Genome U34 Set AB011369_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21913945 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359443 - -
          RatRef-12 Expression BeadChip ILMN_55616 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline