Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573091_m1
          See other KCNT1 GE Assays ›
          Gene Symbol:
          Kcnt1
          Gene Name:
          potassium channel, sodium-activated subfamily T, member 1
          Gene Aliases:
          Slack, rSlo2
          Chromosome Location:
          Chr.3: 3310753 - 3364411 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573091_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021853.1 NP_068625.1 4 - 5 377 102
          XM_006233697.2 XP_006233759.1 4 - 5 685 102
          XM_006233698.2 XP_006233760.1 4 - 5 685 102
          XM_006233699.2 XP_006233761.1 4 - 5 685 102
          XM_006233700.2 XP_006233762.1 3 - 4 643 102
          XM_006233701.2 XP_006233763.1 4 - 5 685 102
          XM_006233702.2 XP_006233764.1 3 - 4 643 102
          XM_006233703.2 XP_006233765.1 4 - 5 685 102
          XM_006233704.2 XP_006233766.1 3 - 4 565 102
          XM_006233705.2 XP_006233767.1 3 - 4 562 102
          XM_006233706.2 XP_006233768.1 4 - 5 685 102
          XM_006233707.2 XP_006233769.1 3 - 4 560 102
          XM_008761624.1 XP_008759846.1 4 - 5 675 102
          GenBank mRNA AF089730.1 - 4 - 5 377 102
          AY884213.1 - 3 - 4 275 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000082.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnt1

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          potassium ion transmembrane transport
          voltage-gated potassium channel activity
          potassium channel activity
          calcium-activated potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003024 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003024 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72826 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834383 - -
          Affymetrix Rat Gene 1.0 ST Array 10834384 - -
          Affymetrix Rat Gene 1.0 ST Array 10834387 - -
          Affymetrix Rat Gene 1.0 ST Array 10834381 - -
          Affymetrix Rat Gene 1.0 ST Array 10834391 - -
          Affymetrix Rat Gene 1.0 ST Array 10834401 - -
          Affymetrix Rat Gene 1.0 ST Array 10834394 - -
          Affymetrix Rat Gene 1.0 ST Array 10834398 - -
          Affymetrix Rat Gene 1.0 ST Array 10834402 - -
          Affymetrix Rat Gene 1.0 ST Array 10834382 - -
          Affymetrix Rat Gene 1.0 ST Array 10834393 - -
          Affymetrix Rat Gene 1.0 ST Array 10834375 - -
          Affymetrix Rat Gene 1.0 ST Array 10834404 - -
          Affymetrix Rat Gene 1.0 ST Array 10834388 - -
          Affymetrix Rat Gene 1.0 ST Array 10834405 - -
          Affymetrix Rat Gene 1.0 ST Array 10834380 - -
          Affymetrix Rat Gene 1.0 ST Array 10834390 - -
          Affymetrix Rat Gene 1.0 ST Array 10834397 - -
          Affymetrix Rat Gene 1.0 ST Array 10834396 - -
          Affymetrix Rat Gene 1.0 ST Array 10834386 - -
          Affymetrix Rat Gene 1.0 ST Array 10834377 - -
          Affymetrix Rat Gene 1.0 ST Array 10834378 - -
          Affymetrix Rat Gene 1.0 ST Array 10834379 - -
          Affymetrix Rat Gene 1.0 ST Array 10834403 - -
          Affymetrix Rat Gene 1.0 ST Array 10834395 - -
          Affymetrix Rat Gene 1.0 ST Array 10834400 - -
          Affymetrix Rat Gene 1.0 ST Array 10834399 - -
          Affymetrix Rat Gene 1.0 ST Array 10834385 - -
          Affymetrix Rat Gene 1.0 ST Array 10834392 - -
          Affymetrix Rat Gene 1.0 ST Array 10834389 - -
          Affymetrix Rat Gene 1.0 ST Array 10834376 - -
          Affymetrix Rat Gene 1.1 ST Array 10834373 - -
          Rat Genome 230 2.0 Array 1384138_at - -
          Rat Genome 230 2.0 Array 1379147_PM_at - -
          Rat Genome 230 2.0 Array 1384138_PM_at - -
          Rat Genome 230 2.0 Array 1395102_PM_at - -
          Rat Genome 230 2.0 Array 1369222_at - -
          Rat Genome 230 2.0 Array 1369222_PM_at - -
          Rat Genome 230 2.0 Array 1379147_at - -
          Rat Genome 230 2.0 Array 1395102_at - -
          Affymetrix HT Rat Focus Array 1395102_at - -
          Affymetrix HT Rat Focus Array 1369222_at - -
          Affymetrix HT Rat Focus Array 1379147_PM_at - -
          Affymetrix HT Rat Focus Array 1395102_PM_at - -
          Affymetrix HT Rat Focus Array 1379147_at - -
          Affymetrix HT Rat Focus Array 1369222_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300000796.rn.1 TC0300000082.rn.1 RTA TC
          Rat Expression Set 230 1369222_PM_at - -
          Rat Expression Set 230 1369222_at - -
          Rat Expression Set 230 1395102_PM_at - -
          Rat Expression Set 230 1379147_PM_at - -
          Rat Expression Set 230 1379147_at - -
          Rat Expression Set 230 1395102_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834373 - -
          Rat Genome U34 Set AF089730_at - -
          Rat Genome U34 Set AF089730_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21144010 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362650 - -
          RatRef-12 Expression BeadChip ILMN_52946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0