Search
Search
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021854.1 | NP_068626.1 | 1 - 2 | 104 | 69 | |
| XM_006233843.2 | XP_006233905.1 | 3 - 4 | 558 | 69 | ||
| XM_006233844.2 | XP_006233906.1 | 3 - 4 | 529 | 69 | ||
| XM_006233845.2 | XP_006233907.1 | 3 - 4 | 380 | 69 | ||
| XM_006233846.2 | XP_006233908.1 | 3 - 4 | 2645 | 69 | ||
| XM_006233847.2 | XP_006233909.1 | 3 - 4 | 866 | 69 | ||
| XM_006233849.2 | XP_006233911.1 | 3 - 4 | 558 | 69 | ||
| GenBank mRNA | AB011821.1 | - | 1 - 2 | 104 | 69 | |
Set Membership: |
|