Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573120_m1
          See other CLOCK GE Assays ›
          Gene Symbol:
          Clock
          Gene Name:
          clock circadian regulator
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 34446694 - 34502218 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573120_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clock
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289832.1 NP_001276761.1 12 - 13 1203 112
          NM_021856.2 NP_068628.1 12 - 13 1203 112
          XM_006250869.2 XP_006250931.1 12 - 13 1247 112
          XM_006250870.2 XP_006250932.1 12 - 13 1247 112
          GenBank mRNA AB019258.1 - 12 - 13 1203 112
          AB019259.1 - 12 - 13 1203 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000399.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clock

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          signal transduction
          spermatogenesis
          circadian rhythm
          entrainment of circadian clock
          histone acetylation
          circadian regulation of gene expression
          regulation of hair cycle
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of inflammatory response
          regulation of insulin secretion
          positive regulation of NF-kappaB transcription factor activity
          response to redox state
          cellular response to ionizing radiation
          regulation of type B pancreatic cell development
          negative regulation of glucocorticoid receptor signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          histone acetyltransferase activity
          signal transducer activity
          chromatin DNA binding
          sequence-specific DNA binding
          protein dimerization activity
          E-box binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147863 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075337 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075337 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147863 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075337 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772264 - -
          Affymetrix Rat Gene 1.0 ST Array 10772251 - -
          Affymetrix Rat Gene 1.0 ST Array 10772266 - -
          Affymetrix Rat Gene 1.0 ST Array 10772250 - -
          Affymetrix Rat Gene 1.0 ST Array 10772258 - -
          Affymetrix Rat Gene 1.0 ST Array 10772263 - -
          Affymetrix Rat Gene 1.0 ST Array 10772252 - -
          Affymetrix Rat Gene 1.0 ST Array 10772267 - -
          Affymetrix Rat Gene 1.0 ST Array 10772268 - -
          Affymetrix Rat Gene 1.0 ST Array 10772269 - -
          Affymetrix Rat Gene 1.0 ST Array 10772256 - -
          Affymetrix Rat Gene 1.0 ST Array 10772260 - -
          Affymetrix Rat Gene 1.0 ST Array 10772254 - -
          Affymetrix Rat Gene 1.0 ST Array 10772265 - -
          Affymetrix Rat Gene 1.0 ST Array 10772261 - -
          Affymetrix Rat Gene 1.0 ST Array 10772255 - -
          Affymetrix Rat Gene 1.0 ST Array 10772262 - -
          Affymetrix Rat Gene 1.0 ST Array 10772257 - -
          Affymetrix Rat Gene 1.0 ST Array 10772270 - -
          Affymetrix Rat Gene 1.0 ST Array 10772253 - -
          Affymetrix Rat Gene 1.0 ST Array 10772271 - -
          Affymetrix Rat Gene 1.0 ST Array 10772259 - -
          Affymetrix Rat Gene 1.1 ST Array 10772249 - -
          Rat Genome 230 2.0 Array 1369168_PM_a_at - -
          Rat Genome 230 2.0 Array 1369168_a_at - -
          Affymetrix HT Rat Focus Array 1369168_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369168_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400003148.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003124.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003134.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003126.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003137.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003133.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003138.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003152.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003145.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003130.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003131.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003128.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003153.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003132.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003140.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003143.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003149.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003127.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003129.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003146.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003147.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003135.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003136.rn.1 TC1400000399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003125.rn.1 TC1400000399.rn.1 RTA TC
          Rat Expression Set 230 1369168_PM_a_at - -
          Rat Expression Set 230 1369168_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10772249 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21658211 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353787 - -
          RatRef-12 Expression BeadChip ILMN_48523 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline