Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573376_m1
          See other PIK3R2 GE Assays ›
          Gene Symbol:
          Pik3r2
          Gene Name:
          phosphoinositide-3-kinase, regulatory subunit 2 (beta)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 20416432 - 20424982 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573376_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3r2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022185.2 NP_071521.2 2 - 3 764 113
          XM_008771096.1 XP_008769318.1 1 - 2 1924 113
          GenBank mRNA BC089805.1 - 2 - 3 764 7381613 113
          D64046.1 - 1 - 2 324 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000285.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pik3r2

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          signal transduction
          regulation of autophagy
          phosphatidylinositol 3-kinase signaling
          protein transport
          phosphorylation
          cellular response to insulin stimulus
          response to endoplasmic reticulum stress
          phosphatidylinositol-3-phosphate biosynthetic process
          positive regulation of transcription factor import into nucleus
          regulation of phosphatidylinositol 3-kinase activity
          positive regulation of transcription from RNA polymerase II promoter
          transcription factor binding
          1-phosphatidylinositol-3-kinase activity
          protein phosphatase binding
          receptor tyrosine kinase binding
          phosphatidylinositol 3-kinase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P158216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927247 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P158216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P158216 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P158216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P158216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12150 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700766143 - -
          Agilent Rat cDNA Microarray (G4105A) 700766143 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787481 - -
          Affymetrix Rat Gene 1.0 ST Array 10787479 - -
          Affymetrix Rat Gene 1.0 ST Array 10787476 - -
          Affymetrix Rat Gene 1.0 ST Array 10787484 - -
          Affymetrix Rat Gene 1.0 ST Array 10787480 - -
          Affymetrix Rat Gene 1.0 ST Array 10787488 - -
          Affymetrix Rat Gene 1.0 ST Array 10787475 - -
          Affymetrix Rat Gene 1.0 ST Array 10787487 - -
          Affymetrix Rat Gene 1.0 ST Array 10787474 - -
          Affymetrix Rat Gene 1.0 ST Array 10787486 - -
          Affymetrix Rat Gene 1.0 ST Array 10787478 - -
          Affymetrix Rat Gene 1.0 ST Array 10787483 - -
          Affymetrix Rat Gene 1.0 ST Array 10787473 - -
          Affymetrix Rat Gene 1.0 ST Array 10787489 - -
          Affymetrix Rat Gene 1.0 ST Array 10787485 - -
          Affymetrix Rat Gene 1.0 ST Array 10787482 - -
          Affymetrix Rat Gene 1.0 ST Array 10787477 - -
          Affymetrix Rat Gene 1.0 ST Array 10787490 - -
          Affymetrix Rat Gene 1.1 ST Array 10787472 - -
          Rat Genome 230 2.0 Array 1376190_PM_at - -
          Rat Genome 230 2.0 Array 1370100_PM_at - -
          Rat Genome 230 2.0 Array 1376190_at - -
          Rat Genome 230 2.0 Array 1370100_at - -
          Affymetrix HT Rat Focus Array 1376190_at - -
          Affymetrix HT Rat Focus Array 1370100_at - -
          Affymetrix HT Rat Focus Array 1376190_PM_at - -
          Affymetrix HT Rat Focus Array 1370100_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600002869.rn.1 TC1600000285.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600002868.rn.1 TC1600000285.rn.1 RTA TC
          Rat Expression Set 230 1376190_PM_at - -
          Rat Expression Set 230 1376190_at - -
          Rat Expression Set 230 1370100_PM_at - -
          Rat Expression Set 230 1370100_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787472 - -
          Rat Genome U34 Set D64046_at - -
          Rat Genome U34 Set D64046_g_at - -
          Rat Genome U34 Set rc_AI103391_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21402129 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650295 - -
          RatRef-12 Expression BeadChip ILMN_50418 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline