Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573408_m1
          See other HTR3B GE Assays ›
          Gene Symbol:
          Htr3b
          Gene Name:
          5-hydroxytryptamine (serotonin) receptor 3B, ionotropic
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 53248705 - 53277859 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htr3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022189.1 NP_071525.1 2 - 3 235 69
          GenBank mRNA AF155044.1 - 2 - 3 235 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002491.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htr3b

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          serotonin receptor signaling pathway
          ion transmembrane transport
          cellular response to growth factor stimulus
          cation transmembrane transport
          G-protein coupled serotonin receptor activity
          extracellular ligand-gated ion channel activity
          serotonin-activated cation-selective channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89615 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014080 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014080 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P478850 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12153 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917136 - -
          Affymetrix Rat Gene 1.0 ST Array 10917141 - -
          Affymetrix Rat Gene 1.0 ST Array 10917139 - -
          Affymetrix Rat Gene 1.0 ST Array 10917137 - -
          Affymetrix Rat Gene 1.0 ST Array 10917144 - -
          Affymetrix Rat Gene 1.0 ST Array 10917140 - -
          Affymetrix Rat Gene 1.0 ST Array 10917143 - -
          Affymetrix Rat Gene 1.0 ST Array 10917138 - -
          Affymetrix Rat Gene 1.0 ST Array 10917142 - -
          Affymetrix Rat Gene 1.1 ST Array 10917135 - -
          Rat Genome 230 2.0 Array 1387602_PM_a_at - -
          Rat Genome 230 2.0 Array 1387602_a_at - -
          Rat Genome 230 2.0 Array 1371077_PM_at - -
          Rat Genome 230 2.0 Array 1371077_at - -
          Affymetrix HT Rat Focus Array 1371077_at - -
          Affymetrix HT Rat Focus Array 1371077_PM_at - -
          Affymetrix HT Rat Focus Array 1387602_a_at - -
          Affymetrix HT Rat Focus Array 1387602_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0800020015.rn.1 TC0800002491.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020011.rn.1 TC0800002491.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020012.rn.1 TC0800002491.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020013.rn.1 TC0800002491.rn.1 RTA TC
          Rat Expression Set 230 1371077_at - -
          Rat Expression Set 230 1387602_a_at - -
          Rat Expression Set 230 1387602_PM_a_at - -
          Rat Expression Set 230 1371077_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917135 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21934142 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356352 - -
          RatRef-12 Expression BeadChip ILMN_48155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline