Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573551_m1
          See other UNC5B GE Assays ›
          Gene Symbol:
          Unc5b
          Gene Name:
          unc-5 homolog B (C. elegans)
          Gene Aliases:
          Unc5h2
          Chromosome Location:
          Chr.20: 30339680 - 30406590 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573551_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Unc5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022207.1 NP_071543.1 16 - 17 2668 70
          XM_006256423.2 XP_006256485.1 15 - 16 3031 70
          GenBank mRNA U87306.1 - 16 - 17 2668 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001533.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Unc5b

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          signal transduction
          axon guidance
          positive regulation of phosphatidylinositol 3-kinase signaling
          anterior/posterior axon guidance
          netrin-activated signaling pathway
          negative regulation of neuron apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          netrin receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134727 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12160 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12160 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12160 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22856 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833016 - -
          Affymetrix Rat Gene 1.0 ST Array 10833023 - -
          Affymetrix Rat Gene 1.0 ST Array 10833020 - -
          Affymetrix Rat Gene 1.0 ST Array 10833018 - -
          Affymetrix Rat Gene 1.0 ST Array 10833015 - -
          Affymetrix Rat Gene 1.0 ST Array 10833024 - -
          Affymetrix Rat Gene 1.0 ST Array 10833027 - -
          Affymetrix Rat Gene 1.0 ST Array 10833026 - -
          Affymetrix Rat Gene 1.0 ST Array 10833021 - -
          Affymetrix Rat Gene 1.0 ST Array 10833017 - -
          Affymetrix Rat Gene 1.0 ST Array 10833019 - -
          Affymetrix Rat Gene 1.0 ST Array 10833022 - -
          Affymetrix Rat Gene 1.0 ST Array 10833025 - -
          Affymetrix Rat Gene 1.0 ST Array 10833028 - -
          Affymetrix Rat Gene 1.0 ST Array 10833014 - -
          Affymetrix Rat Gene 1.0 ST Array 10833030 - -
          Affymetrix Rat Gene 1.0 ST Array 10833029 - -
          Affymetrix Rat Gene 1.1 ST Array 10833013 - -
          Rat Genome 230 2.0 Array 1369235_PM_at - -
          Rat Genome 230 2.0 Array 1369235_at - -
          Rat Genome 230 2.0 Array 1393799_at - -
          Rat Genome 230 2.0 Array 1393799_PM_at - -
          Affymetrix HT Rat Focus Array 1369235_at - -
          Affymetrix HT Rat Focus Array 1393799_at - -
          Affymetrix HT Rat Focus Array 1369235_PM_at - -
          Affymetrix HT Rat Focus Array 1393799_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000012460.rn.1 TC2000001533.rn.1 RTA TC
          Rat Expression Set 230 1369235_at - -
          Rat Expression Set 230 1369235_PM_at - -
          Rat Expression Set 230 1393799_PM_at - -
          Rat Expression Set 230 1393799_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833013 - -
          Rat Genome U34 Set U87306_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21987821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360026 - -
          RatRef-12 Expression BeadChip ILMN_67455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline