Search
Search
| Gene Symbol: Cmklr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022218.2 | NP_071554.2 | 2 | 738 | 80 | |
| XM_006249505.2 | XP_006249567.1 | 3 | 1111 | 80 | ||
| XM_006249506.2 | XP_006249568.1 | 3 | 790 | 80 | ||
| XM_006249507.2 | XP_006249569.1 | 3 | 1437 | 80 | ||
| XM_006249508.2 | XP_006249570.1 | 3 | 1054 | 80 | ||
| XM_006249509.2 | XP_006249571.1 | 3 | 954 | 80 | ||
| GenBank mRNA | AJ002745.1 | - | 1 | 645 | 80 | |
Set Membership: |
|