Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573643_m1
          See other GULO GE Assays ›
          Gene Symbol:
          Gulo
          Gene Name:
          gulonolactone (L-) oxidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 42671206 - 42693711 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573643_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gulo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022220.2 NP_071556.2 8 - 9 825 82
          XM_006252146.2 XP_006252208.1 8 - 9 966 82
          GenBank mRNA BC089803.1 - 8 - 9 825 7324344 82
          FQ219140.1 - 8 - 9 850 82
          FQ219632.1 - 8 - 9 933 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001853.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gulo

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-ascorbic acid biosynthetic process
          oxidation-reduction process
          D-arabinono-1,4-lactone oxidase activity
          UDP-N-acetylmuramate dehydrogenase activity
          L-gulonolactone oxidase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15556 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15556 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15556 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784589 - -
          Affymetrix Rat Gene 1.0 ST Array 10784594 - -
          Affymetrix Rat Gene 1.0 ST Array 10784588 - -
          Affymetrix Rat Gene 1.0 ST Array 10784591 - -
          Affymetrix Rat Gene 1.0 ST Array 10784597 - -
          Affymetrix Rat Gene 1.0 ST Array 10784586 - -
          Affymetrix Rat Gene 1.0 ST Array 10784587 - -
          Affymetrix Rat Gene 1.0 ST Array 10784595 - -
          Affymetrix Rat Gene 1.0 ST Array 10784592 - -
          Affymetrix Rat Gene 1.0 ST Array 10784593 - -
          Affymetrix Rat Gene 1.0 ST Array 10784596 - -
          Affymetrix Rat Gene 1.0 ST Array 10784590 - -
          Affymetrix Rat Gene 1.1 ST Array 10784585 - -
          Rat Genome 230 2.0 Array 1369837_PM_at - -
          Rat Genome 230 2.0 Array 1387725_PM_at - -
          Rat Genome 230 2.0 Array 1387725_at - -
          Rat Genome 230 2.0 Array 1369837_at - -
          Affymetrix HT Rat Focus Array 1387725_at - -
          Affymetrix HT Rat Focus Array 1369837_PM_at - -
          Affymetrix HT Rat Focus Array 1369837_at - -
          Affymetrix HT Rat Focus Array 1387725_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500012415.rn.1 TC1500001853.rn.1 RTA TC
          Rat Expression Set 230 1369837_PM_at - -
          Rat Expression Set 230 1369837_at - -
          Rat Expression Set 230 1387725_at - -
          Rat Expression Set 230 1387725_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784585 - -
          Rat Genome U34 Set D14564cds_s_at - -
          Rat Genome U34 Set rc_AI070903_at - -
          Rat Genome U34 Set D14564cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21609132 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650104 - -
          RatRef-12 Expression BeadChip ILMN_58251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0