Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573797_m1
          See other CYB5A GE Assays ›
          Gene Symbol:
          Cyb5a
          Gene Name:
          cytochrome b5 type A (microsomal)
          Gene Aliases:
          Cyb5
          Chromosome Location:
          Chr.18: 81694818 - 81726821 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573797_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022245.1 NP_071581.1 3 - 4 357 110
          GenBank mRNA AF007108.1 - 3 - 4 320 110
          BC086945.1 - 3 - 4 351 7322141 110
          D13205.1 - 3 - 4 357 110
          FQ209861.1 - 3 - 4 348 110
          FQ210790.1 - 3 - 4 345 110
          FQ224354.1 - 3 - 4 348 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000957.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb5a

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to cadmium ion
          oxidation-reduction process
          molecular_function
          electron carrier activity
          enzyme binding
          heme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P374586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67130 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374586 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P374586 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50455 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80899 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803024 - -
          Affymetrix Rat Gene 1.0 ST Array 10803018 - -
          Affymetrix Rat Gene 1.0 ST Array 10803022 - -
          Affymetrix Rat Gene 1.0 ST Array 10803020 - -
          Affymetrix Rat Gene 1.0 ST Array 10803021 - -
          Affymetrix Rat Gene 1.0 ST Array 10803023 - -
          Affymetrix Rat Gene 1.0 ST Array 10800469 - -
          Affymetrix Rat Gene 1.0 ST Array 10803019 - -
          Affymetrix Rat Gene 1.1 ST Array 10800468 - -
          Affymetrix Rat Gene 1.1 ST Array 10803017 - -
          Rat Genome 230 2.0 Array 1386904_a_at - -
          Rat Genome 230 2.0 Array 1386904_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386904_a_at - -
          Affymetrix HT Rat Focus Array 1386904_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1800006768.rn.1 TC1800000957.rn.1 RTA TC
          Rat Expression Set 230 1386904_a_at - -
          Rat Expression Set 230 1386904_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803017 - -
          Rat Genome U34 Set rc_AA945054_s_at - -
          Rat Genome U34 Set AF007107_s_at - -
          Rat Genome U34 Set rc_AA817685_at - -
          Rat Genome U34 Set AF007107_s_at - -
          Rat Genome U34 Set rc_AA945054_s_at - -
          Rat Genome U34 Set rc_AA817685_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21102067 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361209 - -
          RatRef-12 Expression BeadChip ILMN_61761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline