Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573873_s1
          See other GPR85 GE Assays ›
          Gene Symbol:
          Gpr85
          Gene Name:
          G protein-coupled receptor 85
          Gene Aliases:
          Srep2
          Chromosome Location:
          Chr.4: 40377705 - 40387150 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573873_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022254.3 NP_071590.1 2 1308 89
          XM_006236124.2 XP_006236186.1 4 2247 89
          XM_006236126.2 XP_006236188.1 3 2123 89
          XM_008762724.1 XP_008760946.1 3 2038 89
          GenBank mRNA AB040803.1 - 1 1046 89
          AF203907.1 - 1 1070 89
          BC087727.1 - 2 1308 7320496 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr85

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P221990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P221990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P221990 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P221990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P221990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861127 - -
          Affymetrix Rat Gene 1.0 ST Array 10861126 - -
          Affymetrix Rat Gene 1.0 ST Array 10861125 - -
          Affymetrix Rat Gene 1.0 ST Array 10861128 - -
          Affymetrix Rat Gene 1.0 ST Array 10861129 - -
          Affymetrix Rat Gene 1.1 ST Array 10861124 - -
          Rat Genome 230 2.0 Array 1387666_at - -
          Rat Genome 230 2.0 Array 1387666_PM_at - -
          Affymetrix HT Rat Focus Array 1387666_at - -
          Affymetrix HT Rat Focus Array 1387666_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400017639.rn.1 TC0400002477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017638.rn.1 TC0400002477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017640.rn.1 TC0400002477.rn.1 RTA TC
          Rat Expression Set 230 1387666_at - -
          Rat Expression Set 230 1387666_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10861124 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21669692 - -
          ABI Rat Genome Survey Microarray 22272499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371834 - -
          RatRef-12 Expression BeadChip ILMN_58554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline