Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573919_m1
          See other BSPRY GE Assays ›
          Gene Symbol:
          Bspry
          Gene Name:
          B-box and SPRY domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 78334284 - 78356462 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573919_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bspry
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022261.2 NP_071597.2 4 - 5 737 106
          GenBank mRNA AF245225.1 - 2 - 3 264 106
          BC062051.1 - 4 - 5 685 106
          BC095901.1 - 4 - 5 388 106
          BC111380.1 - 3 - 4 291 106
          BC166587.1 - 4 - 5 737 7937509 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000757.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bspry

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          protein binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137461 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126183 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137468 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137461 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137468 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030747 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P347778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53389 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10869334 - -
          Affymetrix Rat Gene 1.0 ST Array 10869333 - -
          Affymetrix Rat Gene 1.0 ST Array 10869337 - -
          Affymetrix Rat Gene 1.0 ST Array 10869336 - -
          Affymetrix Rat Gene 1.0 ST Array 10869338 - -
          Affymetrix Rat Gene 1.0 ST Array 10869335 - -
          Affymetrix Rat Gene 1.0 ST Array 10869332 - -
          Affymetrix Rat Gene 1.1 ST Array 10869331 - -
          Rat Genome 230 2.0 Array 1387629_PM_at - -
          Rat Genome 230 2.0 Array 1379366_a_at - -
          Rat Genome 230 2.0 Array 1379366_PM_a_at - -
          Rat Genome 230 2.0 Array 1387629_at - -
          Affymetrix HT Rat Focus Array 1387629_at - -
          Affymetrix HT Rat Focus Array 1379366_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387629_PM_at - -
          Affymetrix HT Rat Focus Array 1379366_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500005014.rn.1 TC0500000757.rn.1 RTA TC
          Rat Expression Set 230 1387629_at - -
          Rat Expression Set 230 1387629_PM_at - -
          Rat Expression Set 230 1379366_a_at - -
          Rat Expression Set 230 1379366_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10869331 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21664390 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366975 - -
          RatRef-12 Expression BeadChip ILMN_50665 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline