Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574015_m1
          See other NMUR2 GE Assays ›
          Gene Symbol:
          Nmur2
          Gene Name:
          neuromedin U receptor 2
          Gene Aliases:
          FM-4, Nmu2r
          Chromosome Location:
          Chr.10: 41392663 - 41408518 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574015_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nmur2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022275.3 NP_071611.3 3 - 4 1039 63
          GenBank mRNA AB041229.1 - 3 - 4 1039 63
          AF242875.1 - 3 - 4 913 63
          CB556410.1 - 2 - 3 210 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002835.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nmur2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          reduction of food intake in response to dietary excess
          calcium ion transport
          chloride transport
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          chemical synaptic transmission
          central nervous system development
          grooming behavior
          activation of phospholipase A2 activity by calcium-mediated signaling
          inositol phosphate-mediated signaling
          response to pain
          arachidonic acid secretion
          chloride transmembrane transport
          neuromedin U receptor activity
          intracellular calcium activated chloride channel activity
          GTP binding
          neuropeptide receptor activity
          neuromedin U binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164565 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164565 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742840 - -
          Affymetrix Rat Gene 1.0 ST Array 10742836 - -
          Affymetrix Rat Gene 1.0 ST Array 10742839 - -
          Affymetrix Rat Gene 1.0 ST Array 10742837 - -
          Affymetrix Rat Gene 1.0 ST Array 10742838 - -
          Affymetrix Rat Gene 1.1 ST Array 10742835 - -
          Rat Genome 230 2.0 Array 1387332_at - -
          Rat Genome 230 2.0 Array 1387332_PM_at - -
          Affymetrix HT Rat Focus Array 1387332_PM_at - -
          Affymetrix HT Rat Focus Array 1387332_at - -
          Affymetrix Rat Clariom S Assay PSR1000024911.rn.1 TC1000002835.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024912.rn.1 TC1000002835.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024913.rn.1 TC1000002835.rn.1 RTA TC
          Rat Expression Set 230 1387332_at - -
          Rat Expression Set 230 1387332_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742835 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21378974 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373139 - -
          RatRef-12 Expression BeadChip ILMN_58149 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0