Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574125_g1
          See other AIF1 GE Assays ›
          Gene Symbol:
          Aif1
          Gene Name:
          allograft inflammatory factor 1
          Gene Aliases:
          BART-1, Bart1, iba1, mrf-1
          Chromosome Location:
          Chr.20: 5161350 - 5167180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574125_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aif1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017196.3 NP_058892.1 5 - 6 451 126
          XM_006256061.2 XP_006256123.1 4 - 5 825 126
          XM_006256062.2 XP_006256124.1 5 - 6 802 126
          XM_006256063.2 XP_006256125.1 5 - 6 765 126
          XM_006256065.2 XP_006256127.1 7 - 8 601 126
          GenBank mRNA AB000818.1 - 5 - 6 449 126
          BC081822.1 - 3 - 4 1298 126
          D82069.1 - 7 - 8 606 126
          FM053942.1 - 5 - 6 438 126
          FQ231499.1 - 5 - 6 449 126
          U17919.1 - 5 - 6 423 126
          U33471.1 - 3 - 4 1475 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001109.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aif1

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          phagocytosis, engulfment
          inflammatory response
          negative regulation of gene expression
          positive regulation of muscle hyperplasia
          Rac protein signal transduction
          actin filament polymerization
          positive regulation of cell migration
          cellular response to extracellular stimulus
          cellular response to hormone stimulus
          response to cytokine
          positive regulation of T cell proliferation
          macrophage activation
          negative regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          response to axon injury
          actin filament bundle assembly
          response to glucocorticoid
          response to electrical stimulus
          cellular response to morphine
          cellular response to interferon-gamma
          cellular response to hydroperoxide
          negative regulation of smooth muscle cell chemotaxis
          positive regulation of smooth muscle cell chemotaxis
          positive regulation of monocyte chemotaxis
          ruffle assembly
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of T cell migration
          calcium ion binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047714 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047714 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047714 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130983 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047714 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130983 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421534 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700546891 - -
          Agilent Rat cDNA Microarray (G4105A) 700546891 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828042 - -
          Affymetrix Rat Gene 1.0 ST Array 10828036 - -
          Affymetrix Rat Gene 1.0 ST Array 10828041 - -
          Affymetrix Rat Gene 1.0 ST Array 10828035 - -
          Affymetrix Rat Gene 1.0 ST Array 10828039 - -
          Affymetrix Rat Gene 1.0 ST Array 10828038 - -
          Affymetrix Rat Gene 1.0 ST Array 10828044 - -
          Affymetrix Rat Gene 1.0 ST Array 10828043 - -
          Affymetrix Rat Gene 1.0 ST Array 10828033 - -
          Affymetrix Rat Gene 1.0 ST Array 10828045 - -
          Affymetrix Rat Gene 1.0 ST Array 10828037 - -
          Affymetrix Rat Gene 1.0 ST Array 10828034 - -
          Affymetrix Rat Gene 1.0 ST Array 10828040 - -
          Affymetrix Rat Gene 1.1 ST Array 10828032 - -
          Rat Genome 230 2.0 Array 1369489_PM_at - -
          Rat Genome 230 2.0 Array 1369489_at - -
          Rat Genome 230 2.0 Array 1368558_PM_s_at - -
          Rat Genome 230 2.0 Array 1368558_s_at - -
          Affymetrix HT Rat Focus Array 1369489_at - -
          Affymetrix HT Rat Focus Array 1368558_s_at - -
          Affymetrix HT Rat Focus Array 1368558_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369489_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000009339.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009338.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009332.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009343.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009334.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009333.rn.1 TC2000001109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009336.rn.1 TC2000001109.rn.1 RTA TC
          Rat Expression Set 230 1369489_PM_at - -
          Rat Expression Set 230 1368558_s_at - -
          Rat Expression Set 230 1368558_PM_s_at - -
          Rat Expression Set 230 1369489_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828032 - -
          Rat Genome U34 Set U17919_s_at - -
          Rat Genome U34 Set U10894_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21312781 - -
          ABI Rat Genome Survey Microarray 21565898 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359219 - -
          RatRef-12 Expression BeadChip ILMN_47707 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline