Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574354_m1
          See other PAFAH1B2 GE Assays ›
          Gene Symbol:
          Pafah1b2
          Gene Name:
          platelet-activating factor acetylhydrolase 1b, catalytic subunit 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 50256969 - 50310044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pafah1b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022387.3 NP_071782.1 5 - 6 536 103
          XM_006242957.2 XP_006243019.1 5 - 6 1002 103
          XM_006242958.2 XP_006243020.1 5 - 6 535 103
          XM_008766172.1 XP_008764394.1 5 - 6 766 103
          GenBank mRNA AF016048.1 - 5 - 6 499 103
          BC081714.1 - 5 - 6 536 7108923 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002452.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pafah1b2

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          brain development
          lipid catabolic process
          positive regulation of macroautophagy
          1-alkyl-2-acetylglycerophosphocholine esterase activity
          protein binding
          protein homodimerization activity
          protein heterodimerization activity
          platelet-activating factor acetyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P209459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75865 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P209459 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P209459 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P209459 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P209459 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12216 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700768754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66570 - -
          Agilent Rat cDNA Microarray (G4105A) 700768754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917075 - -
          Affymetrix Rat Gene 1.0 ST Array 10917074 - -
          Affymetrix Rat Gene 1.0 ST Array 10917073 - -
          Affymetrix Rat Gene 1.0 ST Array 10917070 - -
          Affymetrix Rat Gene 1.0 ST Array 10917071 - -
          Affymetrix Rat Gene 1.0 ST Array 10917072 - -
          Affymetrix Rat Gene 1.1 ST Array 10917069 - -
          Rat Genome 230 2.0 Array 1369641_PM_at - -
          Rat Genome 230 2.0 Array 1369642_PM_at - -
          Rat Genome 230 2.0 Array 1369642_at - -
          Rat Genome 230 2.0 Array 1369641_at - -
          Affymetrix HT Rat Focus Array 1369642_PM_at - -
          Affymetrix HT Rat Focus Array 1369642_at - -
          Affymetrix HT Rat Focus Array 1369641_PM_at - -
          Affymetrix HT Rat Focus Array 1369641_at - -
          Affymetrix Rat Clariom S Assay PSR0800019763.rn.1 TC0800002452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019759.rn.1 TC0800002452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019760.rn.1 TC0800002452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019762.rn.1 TC0800002452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019761.rn.1 TC0800002452.rn.1 RTA TC
          Rat Expression Set 230 1369641_at - -
          Rat Expression Set 230 1369642_PM_at - -
          Rat Expression Set 230 1369641_PM_at - -
          Rat Expression Set 230 1369642_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917069 - -
          Rat Genome U34 Set rc_AA899935_at - -
          Rat Genome U34 Set rc_AA998172_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21126056 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358552 - -
          RatRef-12 Expression BeadChip ILMN_65657 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline