Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574367_m1
          See other QDPR GE Assays ›
          Gene Symbol:
          Qdpr
          Gene Name:
          quinoid dihydropteridine reductase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 70164682 - 70178284 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574367_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Qdpr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022390.1 NP_071785.1 1 - 2 146 74
          GenBank mRNA BC072536.1 - 1 - 2 159 7103182 74
          FQ213296.1 - 1 - 2 125 74
          FQ213843.1 - 1 - 2 176 74
          J03481.1 - 1 - 2 146 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000706.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Qdpr

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          L-phenylalanine catabolic process
          tetrahydrobiopterin biosynthetic process
          response to aluminum ion
          response to lead ion
          response to glucagon
          cellular response to drug
          oxidation-reduction process
          6,7-dihydropteridine reductase activity
          protein homodimerization activity
          NADPH binding
          NADH binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93777 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12219 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12219 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12219 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700514177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75554 - -
          Agilent Rat cDNA Microarray (G4105A) 700514177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773085 - -
          Affymetrix Rat Gene 1.0 ST Array 10773082 - -
          Affymetrix Rat Gene 1.0 ST Array 10773087 - -
          Affymetrix Rat Gene 1.0 ST Array 10773081 - -
          Affymetrix Rat Gene 1.0 ST Array 10773084 - -
          Affymetrix Rat Gene 1.0 ST Array 10773086 - -
          Affymetrix Rat Gene 1.0 ST Array 10773083 - -
          Affymetrix Rat Gene 1.1 ST Array 10773080 - -
          Rat Genome 230 2.0 Array 1367695_at - -
          Rat Genome 230 2.0 Array 1367695_PM_at - -
          Affymetrix HT Rat Focus Array 1367695_PM_at - -
          Affymetrix HT Rat Focus Array 1367695_at - -
          Affymetrix Rat Clariom S Assay PSR1400004827.rn.1 TC1400000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004829.rn.1 TC1400000706.rn.1 RTA TC
          Rat Expression Set 230 1367695_PM_at - -
          Rat Expression Set 230 1367695_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773080 - -
          Rat Genome U34 Set J03481mRNA_at - -
          Rat Genome U34 Set J03481mRNA_g_at - -
          Rat Genome U34 Set J03481mRNA_at - -
          Rat Genome U34 Set J03481mRNA_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21675165 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348866 - -
          RatRef-12 Expression BeadChip ILMN_62820 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline