Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574440_m1
          See other SLC25A11 GE Assays ›
          Gene Symbol:
          Slc25a11
          Gene Name:
          solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 57265903 - 57268018 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574440_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022398.2 NP_071793.1 1 - 2 91 145
          GenBank mRNA FQ216657.1 - 119 145
          U84727.1 - 1 - 2 91 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003138.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc25a11

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135925 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135925 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P490703 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700527977 - -
          Agilent Rat cDNA Microarray (G4105A) 700527977 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744489 - -
          Affymetrix Rat Gene 1.0 ST Array 10744493 - -
          Affymetrix Rat Gene 1.0 ST Array 10744488 - -
          Affymetrix Rat Gene 1.0 ST Array 10744490 - -
          Affymetrix Rat Gene 1.0 ST Array 10744491 - -
          Affymetrix Rat Gene 1.0 ST Array 10744492 - -
          Affymetrix Rat Gene 1.0 ST Array 10744486 - -
          Affymetrix Rat Gene 1.1 ST Array 10744485 - -
          Rat Genome 230 2.0 Array 1370060_PM_at - -
          Rat Genome 230 2.0 Array 1370060_at - -
          Affymetrix HT Rat Focus Array 1370060_at - -
          Affymetrix HT Rat Focus Array 1370060_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000028184.rn.1 TC1000003138.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000028185.rn.1 TC1000003138.rn.1 RTA TC
          Rat Expression Set 230 1370060_PM_at - -
          Rat Expression Set 230 1370060_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744485 - -
          Rat Genome U34 Set U84727_at - -
          Rat Genome U34 Set rc_AA859975_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20897236 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374082 - -
          RatRef-12 Expression BeadChip ILMN_66823 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline